Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 45991981-45992254 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PSTPIP2
Entrez Gene ID
9050
Gene Name
proline-serine-threonine phosphatase interacting protein 2
Gene Aliases
MAYP
UniGene
Hs.567384
Gene Chromosome Location
Chr.18: 45983536-46072327 on Build GRCh38
Transcripts
NM_024430.3 XM_011526253.2 XM_011526254.1 XM_011526256.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3821484 c.839-1G>A p.? Chr.18: 45991984-45991984 on Build GRCh38
3821485 c.742-1G>A p.? Chr.18: 45991984-45991984 on Build GRCh38
2877657 c.815G>A p.R272H Chr.18: 45992129-45992129 on Build GRCh38
473852 c.742-1G>T p.? Chr.18: 45992203-45992203 on Build GRCh38
2877658 c.814C>T p.R272C Chr.18: 45992130-45992130 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368472351 Chr.18: 45992058-45992058 on Build GRCh38
rs376133117 Chr.18: 45992024-45992024 on Build GRCh38
rs202000424 Chr.18: 45992222-45992222 on Build GRCh38
rs920820626 Chr.18: 45992143-45992143 on Build GRCh38
rs760219270 Chr.18: 45992004-45992004 on Build GRCh38
rs780258142 Chr.18: 45992096-45992096 on Build GRCh38
rs35539925 Chr.18: 45992214-45992215 on Build GRCh38
rs756139486 Chr.18: 45992095-45992095 on Build GRCh38
rs751442746 Chr.18: 45992208-45992208 on Build GRCh38
rs3842406 Chr.18: 45992215-45992216 on Build GRCh38
rs577894341 Chr.18: 45992202-45992202 on Build GRCh38
rs978301552 Chr.18: 45992011-45992011 on Build GRCh38
rs867222458 Chr.18: 45992200-45992200 on Build GRCh38
rs868368205 Chr.18: 45992180-45992180 on Build GRCh38
rs941034597 Chr.18: 45992109-45992109 on Build GRCh38
rs549116567 Chr.18: 45992063-45992063 on Build GRCh38
rs752577917 Chr.18: 45992194-45992194 on Build GRCh38
rs772732819 Chr.18: 45992141-45992141 on Build GRCh38
rs376930622 Chr.18: 45992191-45992191 on Build GRCh38
rs770502198 Chr.18: 45992153-45992153 on Build GRCh38
rs182921288 Chr.18: 45992047-45992047 on Build GRCh38
rs777658018 Chr.18: 45992079-45992079 on Build GRCh38
rs760272448 Chr.18: 45992151-45992151 on Build GRCh38
rs879074430 Chr.18: 45992083-45992083 on Build GRCh38
rs188026409 Chr.18: 45992144-45992144 on Build GRCh38
rs927146526 Chr.18: 45992052-45992052 on Build GRCh38
rs764778090 Chr.18: 45992193-45992193 on Build GRCh38
rs749589861 Chr.18: 45992112-45992112 on Build GRCh38
rs191266131 Chr.18: 45992197-45992197 on Build GRCh38
rs765149024 Chr.18: 45992025-45992025 on Build GRCh38
rs201831132 Chr.18: 45992212-45992212 on Build GRCh38
rs537771128 Chr.18: 45992186-45992186 on Build GRCh38
rs985247783 Chr.18: 45992067-45992067 on Build GRCh38
rs113089939 Chr.18: 45992212-45992213 on Build GRCh38
rs760686151 Chr.18: 45992129-45992129 on Build GRCh38
rs751314460 Chr.18: 45992207-45992208 on Build GRCh38
rs931051226 Chr.18: 45992005-45992005 on Build GRCh38
rs776002307 Chr.18: 45992165-45992165 on Build GRCh38
rs750533418 Chr.18: 45992088-45992088 on Build GRCh38
rs66500811 Chr.18: 45992213-45992214 on Build GRCh38
rs910883149 Chr.18: 45992099-45992099 on Build GRCh38
rs77356397 Chr.18: 45992207-45992207 on Build GRCh38
rs768747363 Chr.18: 45992128-45992128 on Build GRCh38
rs766149108 Chr.18: 45992023-45992023 on Build GRCh38
rs371604789 Chr.18: 45992130-45992130 on Build GRCh38

View Full Product Details