Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 21291905-21292152 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NCAM2
Entrez Gene ID
4685
Gene Name
neural cell adhesion molecule 2
Gene Aliases
NCAM21
UniGene
Hs.473450
Gene Chromosome Location
Chr.21: 20998315-21543324 on Build GRCh38
Transcripts
NM_004540.3 XM_011529575.2 XM_011529576.2 XM_011529579.2 XM_011529580.2 XM_011529581.2 XM_011529582.2 XM_011529585.2 XM_017028353.1 XM_017028354.1 XM_017028355.1 XM_017028356.1 XM_017028357.1 XM_017028358.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5770054 c.579C>G p.N193K Chr.21: 21292126-21292126 on Build GRCh38
1029562 c.514C>A p.Q172K Chr.21: 21292136-21292136 on Build GRCh38
478456 c.485G>C p.R162P Chr.21: 21292107-21292107 on Build GRCh38
5663428 c.559C>A p.R187R Chr.21: 21292106-21292106 on Build GRCh38
5161590 c.564C>T p.F188F Chr.21: 21292111-21292111 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745983097 Chr.21: 21292063-21292063 on Build GRCh38
rs898441273 Chr.21: 21292112-21292112 on Build GRCh38
rs757974090 Chr.21: 21292115-21292115 on Build GRCh38
rs886447969 Chr.21: 21291939-21291939 on Build GRCh38
rs374401078 Chr.21: 21292121-21292121 on Build GRCh38
rs765315095 Chr.21: 21292100-21292100 on Build GRCh38
rs772294530 Chr.21: 21292065-21292065 on Build GRCh38
rs8134502 Chr.21: 21292075-21292075 on Build GRCh38
rs762586053 Chr.21: 21292104-21292104 on Build GRCh38
rs749923007 Chr.21: 21292103-21292103 on Build GRCh38
rs766008654 Chr.21: 21292106-21292106 on Build GRCh38
rs751188340 Chr.21: 21292107-21292107 on Build GRCh38
rs757840167 Chr.21: 21292001-21292001 on Build GRCh38
rs779401813 Chr.21: 21292123-21292123 on Build GRCh38
rs1006198218 Chr.21: 21291957-21291957 on Build GRCh38
rs1038377851 Chr.21: 21292027-21292027 on Build GRCh38
rs760459830 Chr.21: 21292085-21292085 on Build GRCh38
rs370739586 Chr.21: 21292086-21292086 on Build GRCh38
rs553981033 Chr.21: 21292003-21292003 on Build GRCh38
rs886459366 Chr.21: 21292042-21292042 on Build GRCh38
rs750950880 Chr.21: 21292087-21292087 on Build GRCh38
rs776633904 Chr.21: 21292093-21292093 on Build GRCh38
rs1047733311 Chr.21: 21291958-21291958 on Build GRCh38

View Full Product Details