Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 70732355-70732600 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SMAP1
Entrez Gene ID
60682
Gene Name
small ArfGAP 1
Gene Aliases
SMAP-1
UniGene
Hs.485717
Gene Chromosome Location
Chr.6: 70667771-70862015 on Build GRCh38
Transcripts
NM_001044305.2 NM_001281439.1 NM_001281440.1 NM_021940.4 XM_005248760.4 XM_011536045.2 XM_011536046.2 XM_011536047.2 XM_011536048.2 XM_017011170.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3697938 c.162C>T p.I54I Chr.6: 70732421-70732421 on Build GRCh38
5061912 c.190G>T p.G64W Chr.6: 70732449-70732449 on Build GRCh38
1081310 c.182G>T p.R61I Chr.6: 70732441-70732441 on Build GRCh38
4773039 c.145G>T p.G49C Chr.6: 70732404-70732404 on Build GRCh38
3875242 c.195T>C p.V65V Chr.6: 70732454-70732454 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1039326150 Chr.6: 70732564-70732564 on Build GRCh38
rs575613288 Chr.6: 70732469-70732469 on Build GRCh38
rs766620422 Chr.6: 70732391-70732391 on Build GRCh38
rs143442603 Chr.6: 70732475-70732475 on Build GRCh38
rs757279993 Chr.6: 70732481-70732481 on Build GRCh38
rs773659955 Chr.6: 70732541-70732541 on Build GRCh38
rs900110631 Chr.6: 70732569-70732569 on Build GRCh38
rs973947440 Chr.6: 70732409-70732409 on Build GRCh38
rs901405354 Chr.6: 70732526-70732526 on Build GRCh38
rs371402844 Chr.6: 70732404-70732404 on Build GRCh38
rs778505000 Chr.6: 70732487-70732487 on Build GRCh38
rs777004530 Chr.6: 70732531-70732531 on Build GRCh38
rs771547864 Chr.6: 70732508-70732508 on Build GRCh38
rs370455616 Chr.6: 70732523-70732523 on Build GRCh38
rs72923709 Chr.6: 70732522-70732522 on Build GRCh38
rs183002924 Chr.6: 70732385-70732385 on Build GRCh38
rs770290254 Chr.6: 70732538-70732538 on Build GRCh38
rs753784483 Chr.6: 70732476-70732476 on Build GRCh38
rs888420664 Chr.6: 70732427-70732427 on Build GRCh38
rs374707521 Chr.6: 70732499-70732499 on Build GRCh38
rs751791711 Chr.6: 70732455-70732455 on Build GRCh38
rs943743840 Chr.6: 70732543-70732543 on Build GRCh38
rs779743400 Chr.6: 70732509-70732509 on Build GRCh38
rs763078563 Chr.6: 70732558-70732558 on Build GRCh38

View Full Product Details