Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 73096756-73096999 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RBM25
Entrez Gene ID
58517
Gene Name
RNA binding motif protein 25
Gene Aliases
NET52, RED120, RNPC7, S164, Snu71, fSAP94
UniGene
Hs.531106
Gene Chromosome Location
Chr.14: 73058436-73121675 on Build GRCh38
Transcripts
NM_021239.2 XM_011537044.2 XM_011537045.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4052229 c.548C>T p.A183V Chr.14: 73096919-73096919 on Build GRCh38
6197053 c.616A>G p.M206V Chr.14: 73096987-73096987 on Build GRCh38
328339 c.544-7G>A p.? Chr.14: 73096908-73096908 on Build GRCh38
4630074 c.573C>T p.D191D Chr.14: 73096944-73096944 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs527617400 Chr.14: 73096792-73096792 on Build GRCh38
rs759929827 Chr.14: 73096917-73096917 on Build GRCh38
rs113774219 Chr.14: 73096908-73096908 on Build GRCh38
rs772386351 Chr.14: 73096890-73096890 on Build GRCh38
rs35597037 Chr.14: 73096888-73096889 on Build GRCh38
rs1025462897 Chr.14: 73096798-73096798 on Build GRCh38
rs367772590 Chr.14: 73096932-73096932 on Build GRCh38
rs372596323 Chr.14: 73096954-73096954 on Build GRCh38
rs370349915 Chr.14: 73096944-73096944 on Build GRCh38
rs750060631 Chr.14: 73096945-73096945 on Build GRCh38
rs183386377 Chr.14: 73096891-73096891 on Build GRCh38
rs898500380 Chr.14: 73096843-73096843 on Build GRCh38
rs79323036 Chr.14: 73096873-73096873 on Build GRCh38
rs766642426 Chr.14: 73096907-73096907 on Build GRCh38
rs972530701 Chr.14: 73096787-73096787 on Build GRCh38
rs766042570 Chr.14: 73096921-73096921 on Build GRCh38
rs549577756 Chr.14: 73096898-73096898 on Build GRCh38
rs375825544 Chr.14: 73096956-73096956 on Build GRCh38
rs199555076 Chr.14: 73096865-73096865 on Build GRCh38
rs761064277 Chr.14: 73096893-73096893 on Build GRCh38
rs776273843 Chr.14: 73096899-73096903 on Build GRCh38
rs994074818 Chr.14: 73096847-73096849 on Build GRCh38
rs751029903 Chr.14: 73096935-73096935 on Build GRCh38

View Full Product Details