Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 116918984-116919209 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RFX6
Entrez Gene ID
222546
Gene Name
regulatory factor X6
Gene Aliases
MTCHRS, MTFS, RFXDC1, dJ955L16.1
UniGene
Hs.352276
Gene Chromosome Location
Chr.6: 116877213-116932163 on Build GRCh38
Transcripts
NM_173560.3 XM_011535589.1 XM_017010477.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3157552 c.1037T>A p.I346K Chr.6: 116919151-116919151 on Build GRCh38
395315 c.1056T>G p.N352K Chr.6: 116919170-116919170 on Build GRCh38
3619760 c.1084G>A p.E362K Chr.6: 116919198-116919198 on Build GRCh38
3947930 c.1058G>T p.W353L Chr.6: 116919172-116919172 on Build GRCh38
7246467 c.1033G>T p.D345Y Chr.6: 116919147-116919147 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372515802 Chr.6: 116919164-116919164 on Build GRCh38
rs537177486 Chr.6: 116919135-116919135 on Build GRCh38
rs79696244 Chr.6: 116919103-116919103 on Build GRCh38
rs374987661 Chr.6: 116919094-116919094 on Build GRCh38
rs565976277 Chr.6: 116919022-116919022 on Build GRCh38
rs997638177 Chr.6: 116919043-116919043 on Build GRCh38
rs936886731 Chr.6: 116919061-116919061 on Build GRCh38
rs886279678 Chr.6: 116919051-116919051 on Build GRCh38
rs377109583 Chr.6: 116919172-116919172 on Build GRCh38
rs548232028 Chr.6: 116919080-116919080 on Build GRCh38
rs772582701 Chr.6: 116919159-116919159 on Build GRCh38
rs757784880 Chr.6: 116919094-116919095 on Build GRCh38
rs1026390758 Chr.6: 116919050-116919050 on Build GRCh38
rs781032130 Chr.6: 116919093-116919093 on Build GRCh38
rs529682425 Chr.6: 116919048-116919048 on Build GRCh38
rs768641819 Chr.6: 116919087-116919087 on Build GRCh38
rs988386034 Chr.6: 116919078-116919078 on Build GRCh38

View Full Product Details