Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 37372219-37372472 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
QPCT
Entrez Gene ID
25797
Gene Name
glutaminyl-peptide cyclotransferase
Gene Aliases
GCT, QC, sQC
UniGene
Hs.79033
Gene Chromosome Location
Chr.2: 37344610-37373322 on Build GRCh38
Transcripts
NM_012413.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5953896 c.938G>A p.R313K Chr.2: 37372470-37372470 on Build GRCh38
6760954 c.844G>A p.G282S Chr.2: 37372376-37372376 on Build GRCh38
6354833 c.869A>G p.E290G Chr.2: 37372401-37372401 on Build GRCh38
3183707 c.874C>T p.R292W Chr.2: 37372406-37372406 on Build GRCh38
4923714 c.895T>A p.Y299N Chr.2: 37372427-37372427 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774540075 Chr.2: 37372359-37372359 on Build GRCh38
rs922438941 Chr.2: 37372256-37372256 on Build GRCh38
rs769167091 Chr.2: 37372404-37372404 on Build GRCh38
rs374931233 Chr.2: 37372343-37372343 on Build GRCh38
rs754771209 Chr.2: 37372372-37372372 on Build GRCh38
rs762938073 Chr.2: 37372358-37372358 on Build GRCh38
rs769079122 Chr.2: 37372338-37372338 on Build GRCh38
rs557893780 Chr.2: 37372342-37372342 on Build GRCh38
rs766397861 Chr.2: 37372362-37372362 on Build GRCh38
rs761118409 Chr.2: 37372335-37372335 on Build GRCh38
rs987957027 Chr.2: 37372277-37372277 on Build GRCh38
rs777028151 Chr.2: 37372340-37372340 on Build GRCh38
rs760633891 Chr.2: 37372335-37372337 on Build GRCh38
rs112946006 Chr.2: 37372346-37372346 on Build GRCh38
rs748891521 Chr.2: 37372391-37372391 on Build GRCh38
rs763901290 Chr.2: 37372362-37372362 on Build GRCh38
rs759418916 Chr.2: 37372437-37372437 on Build GRCh38
rs756668058 Chr.2: 37372393-37372393 on Build GRCh38
rs773294067 Chr.2: 37372414-37372414 on Build GRCh38
rs955170717 Chr.2: 37372259-37372259 on Build GRCh38
rs774374809 Chr.2: 37372434-37372434 on Build GRCh38
rs780619329 Chr.2: 37372400-37372400 on Build GRCh38
rs946443286 Chr.2: 37372297-37372297 on Build GRCh38
rs192361161 Chr.2: 37372350-37372350 on Build GRCh38
rs762492322 Chr.2: 37372324-37372324 on Build GRCh38
rs767418608 Chr.2: 37372440-37372440 on Build GRCh38
rs964307384 Chr.2: 37372242-37372242 on Build GRCh38
rs760112401 Chr.2: 37372377-37372377 on Build GRCh38
rs777256535 Chr.2: 37372390-37372390 on Build GRCh38
rs766302620 Chr.2: 37372427-37372427 on Build GRCh38
rs751394872 Chr.2: 37372370-37372370 on Build GRCh38
rs115966900 Chr.2: 37372305-37372305 on Build GRCh38
rs763106664 Chr.2: 37372424-37372424 on Build GRCh38
rs370391513 Chr.2: 37372402-37372402 on Build GRCh38
rs775852741 Chr.2: 37372333-37372333 on Build GRCh38
rs1043457953 Chr.2: 37372292-37372292 on Build GRCh38
rs755809921 Chr.2: 37372381-37372381 on Build GRCh38
rs748722633 Chr.2: 37372321-37372321 on Build GRCh38
rs772671681 Chr.2: 37372323-37372323 on Build GRCh38
rs374776642 Chr.2: 37372405-37372405 on Build GRCh38
rs752628499 Chr.2: 37372331-37372333 on Build GRCh38
rs943186182 Chr.2: 37372307-37372307 on Build GRCh38
rs748494284 Chr.2: 37372406-37372406 on Build GRCh38
rs370745161 Chr.2: 37372247-37372247 on Build GRCh38
rs767257515 Chr.2: 37372373-37372373 on Build GRCh38
rs913683596 Chr.2: 37372295-37372295 on Build GRCh38
rs770121520 Chr.2: 37372407-37372407 on Build GRCh38

View Full Product Details