Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 163325892-163326145 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NPY1R
Entrez Gene ID
4886
Gene Name
neuropeptide Y receptor Y1
Gene Aliases
NPY1-R, NPYR
UniGene
Hs.519057
Gene Chromosome Location
Chr.4: 163323962-163344847 on Build GRCh38
Transcripts
NM_000909.5 XM_005263031.3 XM_011532010.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3720342 c.635C>T p.T212I Chr.4: 163325920-163325920 on Build GRCh38
6205925 c.623G>A p.R208K Chr.4: 163325932-163325932 on Build GRCh38
733237 c.540T>G p.T180T Chr.4: 163326015-163326015 on Build GRCh38
5576879 c.641T>C p.L214P Chr.4: 163325914-163325914 on Build GRCh38
3714736 c.515C>T p.P172L Chr.4: 163326040-163326040 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs868334689 Chr.4: 163326014-163326014 on Build GRCh38
rs760387451 Chr.4: 163326054-163326054 on Build GRCh38
rs757037175 Chr.4: 163325983-163325983 on Build GRCh38
rs774512237 Chr.4: 163326078-163326078 on Build GRCh38
rs763046807 Chr.4: 163326049-163326049 on Build GRCh38
rs990965813 Chr.4: 163326024-163326024 on Build GRCh38
rs760750575 Chr.4: 163326103-163326103 on Build GRCh38
rs781606902 Chr.4: 163326015-163326015 on Build GRCh38
rs148479836 Chr.4: 163326118-163326118 on Build GRCh38
rs183843859 Chr.4: 163326119-163326119 on Build GRCh38
rs773209839 Chr.4: 163325936-163325936 on Build GRCh38
rs752715586 Chr.4: 163325946-163325946 on Build GRCh38
rs769959052 Chr.4: 163326044-163326044 on Build GRCh38
rs139898883 Chr.4: 163326020-163326020 on Build GRCh38
rs189884361 Chr.4: 163325967-163325967 on Build GRCh38
rs377231377 Chr.4: 163325982-163325982 on Build GRCh38
rs370483910 Chr.4: 163325966-163325966 on Build GRCh38
rs762084285 Chr.4: 163325926-163325926 on Build GRCh38
rs75430619 Chr.4: 163326048-163326048 on Build GRCh38
rs149471641 Chr.4: 163325938-163325938 on Build GRCh38
rs867102557 Chr.4: 163326035-163326035 on Build GRCh38
rs181222308 Chr.4: 163326006-163326006 on Build GRCh38
rs150935202 Chr.4: 163325997-163325997 on Build GRCh38
rs760625911 Chr.4: 163325954-163325954 on Build GRCh38
rs772998084 Chr.4: 163325943-163325943 on Build GRCh38
rs966733858 Chr.4: 163325988-163325988 on Build GRCh38
rs766321385 Chr.4: 163325944-163325944 on Build GRCh38
rs771049472 Chr.4: 163326060-163326060 on Build GRCh38
rs146250943 Chr.4: 163326088-163326088 on Build GRCh38
rs778596539 Chr.4: 163325990-163325990 on Build GRCh38
rs1003991153 Chr.4: 163325952-163325952 on Build GRCh38
rs770054149 Chr.4: 163325928-163325928 on Build GRCh38
rs370691271 Chr.4: 163326007-163326007 on Build GRCh38

View Full Product Details