Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 100111921-100112194 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TAF6
Entrez Gene ID
6878
Gene Name
TATA-box binding protein associated factor 6
Gene Aliases
MGC:8964, TAF(II)70, TAF(II)80, TAF2E, TAFII-70, TAFII-80, TAFII70, TAFII80, TAFII85
UniGene
Hs.489309
Gene Chromosome Location
Chr.7: 100107070-100119858 on Build GRCh38
Transcripts
NM_001190415.1 NM_005641.3 NM_139315.2 NR_033792.1 XM_006716100.2 XM_006716101.2 XM_006716102.2 XM_011516542.2 XM_011516543.2 XM_017012568.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6887766 c.743C>T p.T248M Chr.7: 100111977-100111977 on Build GRCh38
6796415 c.685G>A p.E229K Chr.7: 100112143-100112143 on Build GRCh38
5408407 c.726C>T p.A242A Chr.7: 100111994-100111994 on Build GRCh38
5710774 c.751G>T p.G251* Chr.7: 100111969-100111969 on Build GRCh38
4836961 c.760C>G p.Q254E Chr.7: 100111960-100111960 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779876829 Chr.7: 100112073-100112073 on Build GRCh38
rs887575177 Chr.7: 100112072-100112072 on Build GRCh38
rs368675185 Chr.7: 100112043-100112043 on Build GRCh38
rs756451961 Chr.7: 100112033-100112033 on Build GRCh38
rs141218023 Chr.7: 100112123-100112123 on Build GRCh38
rs754251564 Chr.7: 100112162-100112162 on Build GRCh38
rs761312376 Chr.7: 100112144-100112144 on Build GRCh38
rs951630370 Chr.7: 100112095-100112095 on Build GRCh38
rs1007946906 Chr.7: 100111977-100111977 on Build GRCh38
rs377046231 Chr.7: 100112005-100112005 on Build GRCh38
rs546587729 Chr.7: 100112071-100112071 on Build GRCh38
rs761683297 Chr.7: 100112109-100112109 on Build GRCh38
rs372560412 Chr.7: 100111947-100111947 on Build GRCh38
rs768172277 Chr.7: 100112026-100112026 on Build GRCh38
rs751745314 Chr.7: 100112007-100112007 on Build GRCh38
rs374459465 Chr.7: 100112028-100112028 on Build GRCh38
rs200465178 Chr.7: 100112150-100112150 on Build GRCh38
rs746906726 Chr.7: 100112085-100112085 on Build GRCh38
rs753153546 Chr.7: 100112138-100112138 on Build GRCh38
rs370612982 Chr.7: 100112108-100112108 on Build GRCh38
rs1027090777 Chr.7: 100112053-100112053 on Build GRCh38
rs778136553 Chr.7: 100112035-100112035 on Build GRCh38
rs772048032 Chr.7: 100111971-100111971 on Build GRCh38
rs768448576 Chr.7: 100111988-100111988 on Build GRCh38
rs568298776 Chr.7: 100112096-100112096 on Build GRCh38
rs759714011 Chr.7: 100112130-100112130 on Build GRCh38
rs1047642633 Chr.7: 100112065-100112065 on Build GRCh38
rs748917815 Chr.7: 100111942-100111942 on Build GRCh38
rs778517561 Chr.7: 100112049-100112049 on Build GRCh38
rs745371251 Chr.7: 100111964-100111964 on Build GRCh38
rs960202769 Chr.7: 100112153-100112153 on Build GRCh38
rs775465965 Chr.7: 100111975-100111975 on Build GRCh38
rs771325786 Chr.7: 100112115-100112115 on Build GRCh38
rs759663025 Chr.7: 100112013-100112013 on Build GRCh38
rs372996109 Chr.7: 100112135-100112135 on Build GRCh38
rs773318947 Chr.7: 100112003-100112003 on Build GRCh38
rs1039457300 Chr.7: 100111993-100111993 on Build GRCh38
rs745424280 Chr.7: 100112064-100112064 on Build GRCh38
rs541472456 Chr.7: 100111954-100111954 on Build GRCh38
rs756962492 Chr.7: 100112044-100112044 on Build GRCh38
rs768340459 Chr.7: 100112104-100112104 on Build GRCh38
rs750627253 Chr.7: 100111976-100111976 on Build GRCh38
rs766700702 Chr.7: 100112006-100112006 on Build GRCh38

View Full Product Details