Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 24321214-24321471 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPECC1L
Entrez Gene ID
23384
Gene Name
sperm antigen with calponin homology and coiled-coil domains 1 like
Gene Aliases
CYTSA, GBBB2, OBLFC1
UniGene
Hs.474384
Gene Chromosome Location
Chr.22: 24270817-24417740 on Build GRCh38
Transcripts
NM_001145468.3 NM_001254732.2 NM_015330.4 NR_103546.1

Gene Symbol
SPECC1L-ADORA2A
Entrez Gene ID
101730217
Gene Name
SPECC1L-ADORA2A readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.474384
Gene Chromosome Location
Chr.22: 24270817-24442360 on Build GRCh38
Transcripts
NM_001145468.3 NM_001254732.2 NM_015330.4 NR_103546.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6454088 c.386G>A p.R129H Chr.22: 24321366-24321366 on Build GRCh38
1032684 c.350A>C p.K117T Chr.22: 24321330-24321330 on Build GRCh38
3310044 c.352G>A p.E118K Chr.22: 24321332-24321332 on Build GRCh38
1178788 c.464G>T p.R155L Chr.22: 24321444-24321444 on Build GRCh38
3310043 c.328C>T p.R110W Chr.22: 24321308-24321308 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201527235 Chr.22: 24321331-24321331 on Build GRCh38
rs773788526 Chr.22: 24321432-24321432 on Build GRCh38
rs780078274 Chr.22: 24321308-24321308 on Build GRCh38
rs936639910 Chr.22: 24321427-24321427 on Build GRCh38
rs140178777 Chr.22: 24321290-24321290 on Build GRCh38
rs575320570 Chr.22: 24321339-24321339 on Build GRCh38
rs200786886 Chr.22: 24321274-24321274 on Build GRCh38
rs776393240 Chr.22: 24321446-24321446 on Build GRCh38
rs199596465 Chr.22: 24321366-24321366 on Build GRCh38
rs768857920 Chr.22: 24321311-24321311 on Build GRCh38
rs200248577 Chr.22: 24321315-24321315 on Build GRCh38
rs201249023 Chr.22: 24321365-24321365 on Build GRCh38
rs149052234 Chr.22: 24321378-24321378 on Build GRCh38
rs774974834 Chr.22: 24321320-24321320 on Build GRCh38
rs746550418 Chr.22: 24321437-24321437 on Build GRCh38
rs766147313 Chr.22: 24321265-24321265 on Build GRCh38
rs746153199 Chr.22: 24321292-24321292 on Build GRCh38
rs200649219 Chr.22: 24321383-24321383 on Build GRCh38
rs754663263 Chr.22: 24321268-24321268 on Build GRCh38
rs201882878 Chr.22: 24321373-24321373 on Build GRCh38
rs769106775 Chr.22: 24321385-24321385 on Build GRCh38
rs199712268 Chr.22: 24321359-24321359 on Build GRCh38
rs201964345 Chr.22: 24321402-24321402 on Build GRCh38
rs769986955 Chr.22: 24321303-24321303 on Build GRCh38
rs867603969 Chr.22: 24321281-24321281 on Build GRCh38
rs973860392 Chr.22: 24321261-24321261 on Build GRCh38
rs200006813 Chr.22: 24321418-24321418 on Build GRCh38
rs1009778712 Chr.22: 24321445-24321445 on Build GRCh38
rs762382337 Chr.22: 24321329-24321329 on Build GRCh38
rs772837068 Chr.22: 24321336-24321336 on Build GRCh38
rs919653895 Chr.22: 24321288-24321288 on Build GRCh38
rs564211771 Chr.22: 24321410-24321410 on Build GRCh38
rs752582237 Chr.22: 24321285-24321285 on Build GRCh38
rs753391082 Chr.22: 24321344-24321344 on Build GRCh38
rs202112942 Chr.22: 24321309-24321309 on Build GRCh38
rs982575776 Chr.22: 24321291-24321291 on Build GRCh38
rs193920978 Chr.22: 24321444-24321444 on Build GRCh38
rs201744911 Chr.22: 24321278-24321278 on Build GRCh38
rs575022840 Chr.22: 24321342-24321342 on Build GRCh38
rs896205068 Chr.22: 24321343-24321343 on Build GRCh38
rs750510935 Chr.22: 24321369-24321369 on Build GRCh38
rs1048759039 Chr.22: 24321312-24321312 on Build GRCh38
rs748723587 Chr.22: 24321411-24321411 on Build GRCh38
rs772722287 Chr.22: 24321417-24321417 on Build GRCh38
rs148203655 Chr.22: 24321443-24321443 on Build GRCh38
rs371842287 Chr.22: 24321351-24321351 on Build GRCh38
rs778940085 Chr.22: 24321284-24321284 on Build GRCh38
rs200192785 Chr.22: 24321249-24321249 on Build GRCh38
rs200462050 Chr.22: 24321360-24321360 on Build GRCh38
rs867491286 Chr.22: 24321431-24321431 on Build GRCh38
rs753681634 Chr.22: 24321266-24321266 on Build GRCh38
rs143037196 Chr.22: 24321442-24321442 on Build GRCh38
rs202099517 Chr.22: 24321242-24321242 on Build GRCh38
rs200724810 Chr.22: 24321372-24321372 on Build GRCh38
rs79310115 Chr.22: 24321272-24321272 on Build GRCh38
rs200283454 Chr.22: 24321280-24321280 on Build GRCh38

View Full Product Details