Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 113920622-113920855 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RBM19
Entrez Gene ID
9904
Gene Name
RNA binding motif protein 19
Gene Aliases
-
UniGene
Hs.7482
Gene Chromosome Location
Chr.12: 113816738-113966371 on Build GRCh38
Transcripts
NM_001146698.1 NM_001146699.1 NM_016196.3 XM_017020280.1 XM_017020281.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4135717 c.2336T>C p.V779A Chr.12: 113920660-113920660 on Build GRCh38
935458 c.2306-3_2306-2insTC p.? Chr.12: 113920692-113920693 on Build GRCh38
1639044 c.2360C>T p.A787V Chr.12: 113920636-113920636 on Build GRCh38
1263905 c.2349G>A p.K783K Chr.12: 113920647-113920647 on Build GRCh38
935457 c.2355G>A p.E785E Chr.12: 113920641-113920641 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs534880914 Chr.12: 113920672-113920672 on Build GRCh38
rs773402216 Chr.12: 113920714-113920714 on Build GRCh38
rs777258139 Chr.12: 113920682-113920682 on Build GRCh38
rs752695046 Chr.12: 113920729-113920729 on Build GRCh38
rs116963633 Chr.12: 113920828-113920828 on Build GRCh38
rs756543615 Chr.12: 113920693-113920694 on Build GRCh38
rs79120696 Chr.12: 113920644-113920644 on Build GRCh38
rs901915936 Chr.12: 113920819-113920819 on Build GRCh38
rs546203528 Chr.12: 113920793-113920793 on Build GRCh38
rs75315956 Chr.12: 113920805-113920805 on Build GRCh38
rs759385346 Chr.12: 113920723-113920723 on Build GRCh38
rs753274889 Chr.12: 113920692-113920693 on Build GRCh38
rs767535416 Chr.12: 113920693-113920696 on Build GRCh38
rs376118231 Chr.12: 113920722-113920722 on Build GRCh38
rs201685232 Chr.12: 113920730-113920730 on Build GRCh38
rs748992034 Chr.12: 113920687-113920687 on Build GRCh38
rs142556388 Chr.12: 113920648-113920648 on Build GRCh38
rs199568921 Chr.12: 113920716-113920716 on Build GRCh38
rs947938887 Chr.12: 113920803-113920803 on Build GRCh38
rs144730831 Chr.12: 113920814-113920814 on Build GRCh38
rs146568369 Chr.12: 113920725-113920725 on Build GRCh38
rs755622048 Chr.12: 113920676-113920676 on Build GRCh38
rs111824990 Chr.12: 113920645-113920645 on Build GRCh38
rs753380341 Chr.12: 113920740-113920740 on Build GRCh38
rs892387095 Chr.12: 113920809-113920809 on Build GRCh38
rs779436539 Chr.12: 113920762-113920762 on Build GRCh38
rs773637304 Chr.12: 113920642-113920642 on Build GRCh38
rs1036483037 Chr.12: 113920817-113920817 on Build GRCh38
rs78057972 Chr.12: 113920761-113920761 on Build GRCh38
rs763637724 Chr.12: 113920739-113920739 on Build GRCh38
rs778393591 Chr.12: 113920689-113920689 on Build GRCh38
rs571031113 Chr.12: 113920777-113920777 on Build GRCh38
rs754688025 Chr.12: 113920652-113920652 on Build GRCh38
rs748662249 Chr.12: 113920710-113920710 on Build GRCh38
rs752515852 Chr.12: 113920675-113920675 on Build GRCh38
rs771215223 Chr.12: 113920720-113920720 on Build GRCh38
rs1053146633 Chr.12: 113920807-113920807 on Build GRCh38
rs200339287 Chr.12: 113920697-113920697 on Build GRCh38
rs945301999 Chr.12: 113920810-113920810 on Build GRCh38
rs562865081 Chr.12: 113920812-113920812 on Build GRCh38
rs142858844 Chr.12: 113920721-113920721 on Build GRCh38
rs747426617 Chr.12: 113920695-113920695 on Build GRCh38
rs112539928 Chr.12: 113920776-113920776 on Build GRCh38
rs777200576 Chr.12: 113920707-113920707 on Build GRCh38
rs767507902 Chr.12: 113920658-113920658 on Build GRCh38
rs763718755 Chr.12: 113920688-113920688 on Build GRCh38
rs59243773 Chr.12: 113920802-113920802 on Build GRCh38
rs372137741 Chr.12: 113920712-113920712 on Build GRCh38

View Full Product Details