Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 47046414-47046639 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM160
Entrez Gene ID
54958
Gene Name
transmembrane protein 160
Gene Aliases
-
UniGene
Hs.105606
Gene Chromosome Location
Chr.19: 47045907-47048624 on Build GRCh38
Transcripts
NM_017854.1 XM_017026917.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
712192 c.279C>A p.D93E Chr.19: 47046615-47046615 on Build GRCh38
6803452 c.287G>A p.R96Q Chr.19: 47046607-47046607 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs756655049 Chr.19: 47046574-47046575 on Build GRCh38
rs971414908 Chr.19: 47046475-47046475 on Build GRCh38
rs993728660 Chr.19: 47046437-47046437 on Build GRCh38
rs952109772 Chr.19: 47046463-47046463 on Build GRCh38
rs751013454 Chr.19: 47046575-47046575 on Build GRCh38
rs924424111 Chr.19: 47046510-47046510 on Build GRCh38
rs370262628 Chr.19: 47046541-47046541 on Build GRCh38
rs762166519 Chr.19: 47046614-47046614 on Build GRCh38
rs550706372 Chr.19: 47046498-47046498 on Build GRCh38
rs752969300 Chr.19: 47046577-47046577 on Build GRCh38
rs917210973 Chr.19: 47046441-47046441 on Build GRCh38
rs370488459 Chr.19: 47046574-47046574 on Build GRCh38
rs780628923 Chr.19: 47046590-47046590 on Build GRCh38
rs934476109 Chr.19: 47046523-47046523 on Build GRCh38
rs764456731 Chr.19: 47046545-47046545 on Build GRCh38
rs991892135 Chr.19: 47046465-47046465 on Build GRCh38
rs890884970 Chr.19: 47046568-47046568 on Build GRCh38
rs780847015 Chr.19: 47046580-47046580 on Build GRCh38
rs762794457 Chr.19: 47046593-47046593 on Build GRCh38
rs932372793 Chr.19: 47046559-47046559 on Build GRCh38
rs117870772 Chr.19: 47046472-47046472 on Build GRCh38
rs1028306006 Chr.19: 47046442-47046442 on Build GRCh38
rs760855236 Chr.19: 47046617-47046617 on Build GRCh38
rs373096639 Chr.19: 47046563-47046563 on Build GRCh38
rs978848393 Chr.19: 47046612-47046612 on Build GRCh38
rs201144157 Chr.19: 47046571-47046571 on Build GRCh38
rs375352283 Chr.19: 47046481-47046481 on Build GRCh38
rs1862499 Chr.19: 47046533-47046533 on Build GRCh38
rs747900007 Chr.19: 47046581-47046581 on Build GRCh38
rs746569691 Chr.19: 47046576-47046576 on Build GRCh38
rs141236807 Chr.19: 47046597-47046597 on Build GRCh38
rs374990760 Chr.19: 47046610-47046610 on Build GRCh38
rs754201890 Chr.19: 47046562-47046562 on Build GRCh38
rs187680020 Chr.19: 47046546-47046546 on Build GRCh38
rs759004653 Chr.19: 47046573-47046573 on Build GRCh38
rs570301672 Chr.19: 47046506-47046506 on Build GRCh38
rs377540942 Chr.19: 47046564-47046564 on Build GRCh38
rs775407111 Chr.19: 47046606-47046606 on Build GRCh38
rs981254007 Chr.19: 47046490-47046490 on Build GRCh38
rs779428828 Chr.19: 47046575-47046575 on Build GRCh38

View Full Product Details