Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 25670661-25670932 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC34A2
Entrez Gene ID
10568
Gene Name
solute carrier family 34 member 2
Gene Aliases
NAPI-3B, NAPI-IIb, NPTIIb
UniGene
Hs.479372
Gene Chromosome Location
Chr.4: 25655813-25678748 on Build GRCh38
Transcripts
NM_001177998.1 NM_001177999.1 NM_006424.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4124190 c.861A>G p.A287A Chr.4: 25670767-25670767 on Build GRCh38
4727144 c.855A>G p.Q285Q Chr.4: 25670761-25670761 on Build GRCh38
1186542 c.835G>A p.D279N Chr.4: 25670741-25670741 on Build GRCh38
7239327 c.864G>C p.M288I Chr.4: 25670770-25670770 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs751883268 Chr.4: 25670837-25670837 on Build GRCh38
rs79937695 Chr.4: 25670747-25670747 on Build GRCh38
rs752197307 Chr.4: 25670830-25670830 on Build GRCh38
rs181267814 Chr.4: 25670776-25670776 on Build GRCh38
rs200876077 Chr.4: 25670693-25670693 on Build GRCh38
rs762076508 Chr.4: 25670864-25670864 on Build GRCh38
rs567904509 Chr.4: 25670860-25670860 on Build GRCh38
rs747355268 Chr.4: 25670748-25670748 on Build GRCh38
rs770465193 Chr.4: 25670855-25670855 on Build GRCh38
rs768813002 Chr.4: 25670785-25670785 on Build GRCh38
rs367869119 Chr.4: 25670782-25670782 on Build GRCh38
rs753759730 Chr.4: 25670809-25670809 on Build GRCh38
rs765458030 Chr.4: 25670725-25670725 on Build GRCh38
rs758026618 Chr.4: 25670738-25670738 on Build GRCh38
rs757711485 Chr.4: 25670838-25670838 on Build GRCh38
rs140631105 Chr.4: 25670773-25670773 on Build GRCh38
rs531268967 Chr.4: 25670797-25670797 on Build GRCh38
rs777320214 Chr.4: 25670834-25670834 on Build GRCh38
rs190736944 Chr.4: 25670718-25670718 on Build GRCh38
rs375862993 Chr.4: 25670811-25670811 on Build GRCh38
rs758019128 Chr.4: 25670833-25670833 on Build GRCh38
rs186725988 Chr.4: 25670861-25670861 on Build GRCh38
rs982150295 Chr.4: 25670745-25670745 on Build GRCh38
rs781729149 Chr.4: 25670845-25670845 on Build GRCh38
rs778448402 Chr.4: 25670723-25670723 on Build GRCh38
rs766244885 Chr.4: 25670808-25670808 on Build GRCh38
rs150930496 Chr.4: 25670771-25670771 on Build GRCh38
rs746374609 Chr.4: 25670853-25670853 on Build GRCh38
rs367580162 Chr.4: 25670721-25670721 on Build GRCh38
rs968407909 Chr.4: 25670807-25670807 on Build GRCh38
rs769981964 Chr.4: 25670774-25670774 on Build GRCh38
rs760496504 Chr.4: 25670804-25670804 on Build GRCh38
rs758895971 Chr.4: 25670765-25670765 on Build GRCh38
rs372023023 Chr.4: 25670784-25670784 on Build GRCh38
rs763143021 Chr.4: 25670692-25670692 on Build GRCh38
rs771522281 Chr.4: 25670769-25670769 on Build GRCh38
rs746488364 Chr.4: 25670858-25670858 on Build GRCh38
rs766328841 Chr.4: 25670700-25670700 on Build GRCh38
rs770704925 Chr.4: 25670873-25670873 on Build GRCh38
rs368385667 Chr.4: 25670856-25670856 on Build GRCh38
rs891203611 Chr.4: 25670684-25670684 on Build GRCh38
rs764692447 Chr.4: 25670829-25670829 on Build GRCh38
rs376213897 Chr.4: 25670699-25670699 on Build GRCh38
rs779522882 Chr.4: 25670867-25670880 on Build GRCh38
rs939657012 Chr.4: 25670896-25670896 on Build GRCh38
rs758102711 Chr.4: 25670743-25670744 on Build GRCh38

View Full Product Details