Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 49090307-49090548 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SNRNP70
Entrez Gene ID
6625
Gene Name
small nuclear ribonucleoprotein U1 subunit 70
Gene Aliases
RNPU1Z, RPU1, SNRP70, Snp1, U1-70K, U170K, U1AP, U1RNP
UniGene
Hs.467097
Gene Chromosome Location
Chr.19: 49085140-49108613 on Build GRCh38
Transcripts
NM_001301069.1 NM_003089.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
135842 c.260A>C p.K87T Chr.19: 49090515-49090515 on Build GRCh38
1395176 c.194A>G p.E65G Chr.19: 49090337-49090337 on Build GRCh38
6842452 c.219A>T p.E73D Chr.19: 49090474-49090474 on Build GRCh38
4955688 c.250A>G p.T84A Chr.19: 49090505-49090505 on Build GRCh38
6375960 c.175C>A p.R59S Chr.19: 49090318-49090318 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1037465945 Chr.19: 49090390-49090390 on Build GRCh38
rs141339800 Chr.19: 49090376-49090376 on Build GRCh38
rs765397185 Chr.19: 49090498-49090498 on Build GRCh38
rs759579766 Chr.19: 49090454-49090454 on Build GRCh38
rs758502375 Chr.19: 49090331-49090331 on Build GRCh38
rs933206474 Chr.19: 49090350-49090350 on Build GRCh38
rs780237357 Chr.19: 49090527-49090527 on Build GRCh38
rs774791055 Chr.19: 49090380-49090381 on Build GRCh38
rs200763700 Chr.19: 49090427-49090427 on Build GRCh38
rs758414387 Chr.19: 49090504-49090504 on Build GRCh38
rs905790459 Chr.19: 49090408-49090408 on Build GRCh38
rs747563092 Chr.19: 49090371-49090371 on Build GRCh38
rs768992931 Chr.19: 49090377-49090378 on Build GRCh38
rs769616941 Chr.19: 49090405-49090405 on Build GRCh38
rs752577951 Chr.19: 49090485-49090485 on Build GRCh38
rs1002781722 Chr.19: 49090458-49090458 on Build GRCh38
rs773019177 Chr.19: 49090417-49090417 on Build GRCh38
rs898559466 Chr.19: 49090416-49090416 on Build GRCh38
rs934304606 Chr.19: 49090370-49090370 on Build GRCh38
rs750647099 Chr.19: 49090500-49090500 on Build GRCh38
rs780470838 Chr.19: 49090368-49090368 on Build GRCh38
rs747720815 Chr.19: 49090401-49090401 on Build GRCh38
rs940027316 Chr.19: 49090388-49090388 on Build GRCh38
rs774101758 Chr.19: 49090449-49090449 on Build GRCh38
rs748505286 Chr.19: 49090445-49090445 on Build GRCh38
rs931403585 Chr.19: 49090433-49090433 on Build GRCh38
rs770573551 Chr.19: 49090434-49090434 on Build GRCh38
rs1055269692 Chr.19: 49090447-49090447 on Build GRCh38
rs374604976 Chr.19: 49090377-49090377 on Build GRCh38
rs767212023 Chr.19: 49090462-49090462 on Build GRCh38
rs762002567 Chr.19: 49090488-49090488 on Build GRCh38

View Full Product Details