Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 173481257-173481530 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRDX6
Entrez Gene ID
9588
Gene Name
peroxiredoxin 6
Gene Aliases
1-Cys, AOP2, HEL-S-128m, NSGPx, PRX, aiPLA2, p29
UniGene
Hs.120
Gene Chromosome Location
Chr.1: 173477347-173488807 on Build GRCh38
Transcripts
NM_004905.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3478469 c.113C>T p.S38F Chr.1: 173481343-173481343 on Build GRCh38
163712 c.194A>T p.N65I Chr.1: 173481424-173481424 on Build GRCh38
6583621 c.224G>T p.S75I Chr.1: 173481454-173481454 on Build GRCh38
5349996 c.238C>G p.L80V Chr.1: 173481468-173481468 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs765762473 Chr.1: 173481454-173481454 on Build GRCh38
rs940362878 Chr.1: 173481315-173481315 on Build GRCh38
rs751080011 Chr.1: 173481471-173481471 on Build GRCh38
rs749075728 Chr.1: 173481499-173481499 on Build GRCh38
rs768723962 Chr.1: 173481382-173481382 on Build GRCh38
rs746524781 Chr.1: 173481332-173481332 on Build GRCh38
rs780743209 Chr.1: 173481298-173481298 on Build GRCh38
rs773452101 Chr.1: 173481416-173481416 on Build GRCh38
rs757997667 Chr.1: 173481309-173481309 on Build GRCh38
rs755610622 Chr.1: 173481453-173481453 on Build GRCh38
rs750807574 Chr.1: 173481290-173481290 on Build GRCh38
rs745706076 Chr.1: 173481343-173481343 on Build GRCh38
rs774323358 Chr.1: 173481504-173481504 on Build GRCh38
rs200221454 Chr.1: 173481316-173481316 on Build GRCh38
rs774357324 Chr.1: 173481401-173481401 on Build GRCh38
rs374525073 Chr.1: 173481299-173481299 on Build GRCh38
rs761935759 Chr.1: 173481409-173481409 on Build GRCh38
rs887178093 Chr.1: 173481389-173481389 on Build GRCh38
rs771701957 Chr.1: 173481341-173481341 on Build GRCh38
rs374364029 Chr.1: 173481475-173481475 on Build GRCh38
rs756522053 Chr.1: 173481293-173481293 on Build GRCh38
rs745477807 Chr.1: 173481484-173481484 on Build GRCh38
rs755918840 Chr.1: 173481488-173481488 on Build GRCh38
rs35052032 Chr.1: 173481318-173481318 on Build GRCh38
rs199713481 Chr.1: 173481345-173481345 on Build GRCh38
rs987370077 Chr.1: 173481285-173481285 on Build GRCh38
rs930355626 Chr.1: 173481458-173481458 on Build GRCh38
rs146176256 Chr.1: 173481352-173481352 on Build GRCh38
rs768528960 Chr.1: 173481503-173481503 on Build GRCh38
rs749163040 Chr.1: 173481365-173481365 on Build GRCh38
rs531436261 Chr.1: 173481494-173481494 on Build GRCh38
rs780743420 Chr.1: 173481477-173481477 on Build GRCh38
rs141501072 Chr.1: 173481344-173481344 on Build GRCh38
rs149535776 Chr.1: 173481410-173481410 on Build GRCh38
rs766798941 Chr.1: 173481440-173481440 on Build GRCh38
rs370697515 Chr.1: 173481447-173481447 on Build GRCh38
rs779754915 Chr.1: 173481492-173481492 on Build GRCh38
rs761151739 Chr.1: 173481426-173481426 on Build GRCh38
rs561003015 Chr.1: 173481336-173481336 on Build GRCh38

View Full Product Details