Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 69744788-69745059 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NOB1
Entrez Gene ID
28987
Gene Name
NIN1/PSMD8 binding protein 1 homolog
Gene Aliases
ART-4, MST158, MSTP158, NOB1P, PSMD8BP1
UniGene
Hs.271695
Gene Chromosome Location
Chr.16: 69741854-69754968 on Build GRCh38
Transcripts
NM_014062.2 NR_074074.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3421121 c.897C>T p.T299T Chr.16: 69744945-69744945 on Build GRCh38
4959269 c.835G>A p.D279N Chr.16: 69745007-69745007 on Build GRCh38
3511309 c.934C>T p.R312C Chr.16: 69744908-69744908 on Build GRCh38
6727536 c.830C>T p.T277M Chr.16: 69745012-69745012 on Build GRCh38
3511310 c.843C>T p.S281S Chr.16: 69744999-69744999 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200093997 Chr.16: 69744944-69744944 on Build GRCh38
rs770525334 Chr.16: 69744859-69744859 on Build GRCh38
rs764369537 Chr.16: 69744882-69744882 on Build GRCh38
rs201985418 Chr.16: 69744884-69744884 on Build GRCh38
rs569105886 Chr.16: 69744961-69744961 on Build GRCh38
rs765288090 Chr.16: 69744906-69744906 on Build GRCh38
rs746682366 Chr.16: 69744858-69744858 on Build GRCh38
rs760833894 Chr.16: 69744985-69744985 on Build GRCh38
rs779451471 Chr.16: 69744969-69744969 on Build GRCh38
rs751491953 Chr.16: 69745004-69745004 on Build GRCh38
rs765697697 Chr.16: 69744820-69744820 on Build GRCh38
rs756136944 Chr.16: 69744901-69744901 on Build GRCh38
rs774798065 Chr.16: 69744883-69744883 on Build GRCh38
rs1025882909 Chr.16: 69744888-69744888 on Build GRCh38
rs775844742 Chr.16: 69744878-69744878 on Build GRCh38
rs767817700 Chr.16: 69744885-69744885 on Build GRCh38
rs375951032 Chr.16: 69744932-69744932 on Build GRCh38
rs774445903 Chr.16: 69744843-69744853 on Build GRCh38
rs372933003 Chr.16: 69744950-69744950 on Build GRCh38
rs763441100 Chr.16: 69744881-69744881 on Build GRCh38
rs201462080 Chr.16: 69744945-69744945 on Build GRCh38
rs536540729 Chr.16: 69744978-69744978 on Build GRCh38
rs777480418 Chr.16: 69744856-69744856 on Build GRCh38
rs746762242 Chr.16: 69744917-69744917 on Build GRCh38
rs777570597 Chr.16: 69744915-69744915 on Build GRCh38
rs761669798 Chr.16: 69745005-69745005 on Build GRCh38
rs139171866 Chr.16: 69745013-69745013 on Build GRCh38
rs143009356 Chr.16: 69744907-69744907 on Build GRCh38
rs750581436 Chr.16: 69744889-69744889 on Build GRCh38
rs751401849 Chr.16: 69744954-69744954 on Build GRCh38
rs748801425 Chr.16: 69744972-69744972 on Build GRCh38
rs779735281 Chr.16: 69745024-69745024 on Build GRCh38
rs778695986 Chr.16: 69744834-69744834 on Build GRCh38
rs530522292 Chr.16: 69745011-69745011 on Build GRCh38
rs774997371 Chr.16: 69744933-69744933 on Build GRCh38
rs769336680 Chr.16: 69744974-69744974 on Build GRCh38
rs369384135 Chr.16: 69744983-69744983 on Build GRCh38
rs1041253548 Chr.16: 69744890-69744890 on Build GRCh38
rs762247376 Chr.16: 69744940-69744940 on Build GRCh38
rs575122853 Chr.16: 69744874-69744874 on Build GRCh38
rs749772355 Chr.16: 69744935-69744935 on Build GRCh38
rs753347977 Chr.16: 69745027-69745027 on Build GRCh38
rs780887579 Chr.16: 69744957-69744957 on Build GRCh38
rs748035004 Chr.16: 69744838-69744838 on Build GRCh38
rs550706516 Chr.16: 69744941-69744941 on Build GRCh38
rs140714914 Chr.16: 69745012-69745012 on Build GRCh38
rs202063989 Chr.16: 69744936-69744936 on Build GRCh38
rs973264118 Chr.16: 69744864-69744864 on Build GRCh38
rs944434138 Chr.16: 69744887-69744887 on Build GRCh38
rs770379216 Chr.16: 69744875-69744875 on Build GRCh38
rs745439543 Chr.16: 69744925-69744925 on Build GRCh38
rs771149481 Chr.16: 69744986-69744986 on Build GRCh38
rs201329814 Chr.16: 69744953-69744953 on Build GRCh38
rs754532902 Chr.16: 69745036-69745036 on Build GRCh38
rs368496320 Chr.16: 69744825-69744825 on Build GRCh38
rs776146294 Chr.16: 69744873-69744873 on Build GRCh38
rs1046581842 Chr.16: 69744974-69744975 on Build GRCh38
rs370129186 Chr.16: 69744921-69744921 on Build GRCh38
rs758126830 Chr.16: 69744849-69744849 on Build GRCh38
rs367807444 Chr.16: 69744951-69744951 on Build GRCh38
rs373158111 Chr.16: 69744997-69744997 on Build GRCh38
rs764073447 Chr.16: 69745003-69745003 on Build GRCh38
rs921404406 Chr.16: 69744898-69744898 on Build GRCh38
rs754984021 Chr.16: 69744826-69744826 on Build GRCh38
rs144374127 Chr.16: 69744980-69744980 on Build GRCh38
rs753909517 Chr.16: 69744947-69744947 on Build GRCh38
rs147206895 Chr.16: 69744939-69744939 on Build GRCh38
rs140632082 Chr.16: 69744967-69744967 on Build GRCh38
rs756940203 Chr.16: 69744919-69744919 on Build GRCh38

View Full Product Details