Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 56820385-56820646 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPINK2
Entrez Gene ID
6691
Gene Name
serine peptidase inhibitor, Kazal type 2
Gene Aliases
HUSI-II
UniGene
Hs.98243
Gene Chromosome Location
Chr.4: 56809860-56822210 on Build GRCh38
Transcripts
NM_001271718.1 NM_001271719.1 NM_001271720.1 NM_001271721.1 NM_001271722.1 NM_021114.3 NR_073417.1 NR_073418.1 NR_073419.1 XM_011534405.1 XM_011534406.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
342488 c.99_99+1GG>TT p.? Chr.4: 56820535-56820536 on Build GRCh38
5789779 c.94A>G p.R32G Chr.4: 56820541-56820541 on Build GRCh38
5734508 c.80_86delTGTTTTC p.L27fs*39 Chr.4: 56820549-56820555 on Build GRCh38
4789640 c.56C>T p.A19V Chr.4: 56820579-56820579 on Build GRCh38
6788970 c.79C>A p.L27M Chr.4: 56820556-56820556 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs911787772 Chr.4: 56820473-56820473 on Build GRCh38
rs812991 Chr.4: 56820491-56820491 on Build GRCh38
rs142611048 Chr.4: 56820538-56820538 on Build GRCh38
rs763514307 Chr.4: 56820610-56820610 on Build GRCh38
rs935744339 Chr.4: 56820533-56820533 on Build GRCh38
rs771038147 Chr.4: 56820587-56820587 on Build GRCh38
rs865923182 Chr.4: 56820472-56820472 on Build GRCh38
rs747189131 Chr.4: 56820492-56820492 on Build GRCh38
rs767259930 Chr.4: 56820540-56820540 on Build GRCh38
rs747068279 Chr.4: 56820585-56820585 on Build GRCh38
rs775849010 Chr.4: 56820517-56820517 on Build GRCh38
rs562310552 Chr.4: 56820589-56820589 on Build GRCh38
rs116611500 Chr.4: 56820563-56820563 on Build GRCh38
rs754935715 Chr.4: 56820579-56820579 on Build GRCh38
rs200773254 Chr.4: 56820559-56820559 on Build GRCh38
rs1046569857 Chr.4: 56820593-56820593 on Build GRCh38
rs750134082 Chr.4: 56820553-56820553 on Build GRCh38
rs763531032 Chr.4: 56820527-56820527 on Build GRCh38
rs765697470 Chr.4: 56820512-56820512 on Build GRCh38
rs376843108 Chr.4: 56820596-56820596 on Build GRCh38
rs763632110 Chr.4: 56820530-56820530 on Build GRCh38
rs943297009 Chr.4: 56820483-56820483 on Build GRCh38
rs376900332 Chr.4: 56820466-56820466 on Build GRCh38
rs112967127 Chr.4: 56820480-56820480 on Build GRCh38
rs114989645 Chr.4: 56820569-56820569 on Build GRCh38
rs557762119 Chr.4: 56820537-56820537 on Build GRCh38
rs771164772 Chr.4: 56820499-56820499 on Build GRCh38
rs776518119 Chr.4: 56820446-56820446 on Build GRCh38
rs760096445 Chr.4: 56820482-56820482 on Build GRCh38
rs1043615188 Chr.4: 56820548-56820548 on Build GRCh38
rs963649067 Chr.4: 56820421-56820421 on Build GRCh38
rs779909326 Chr.4: 56820574-56820574 on Build GRCh38
rs113305946 Chr.4: 56820409-56820409 on Build GRCh38
rs560399027 Chr.4: 56820578-56820578 on Build GRCh38
rs537940835 Chr.4: 56820452-56820452 on Build GRCh38
rs887914888 Chr.4: 56820615-56820615 on Build GRCh38
rs531370610 Chr.4: 56820594-56820594 on Build GRCh38
rs577163578 Chr.4: 56820582-56820582 on Build GRCh38
rs759787819 Chr.4: 56820506-56820506 on Build GRCh38
rs189954381 Chr.4: 56820597-56820597 on Build GRCh38
rs775213221 Chr.4: 56820442-56820442 on Build GRCh38
rs75736293 Chr.4: 56820583-56820583 on Build GRCh38

View Full Product Details