Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 62966566-62966827 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC17A9
Entrez Gene ID
63910
Gene Name
solute carrier family 17 member 9
Gene Aliases
C20orf59, POROK8, VNUT
UniGene
Hs.512686
Gene Chromosome Location
Chr.20: 62952647-62968597 on Build GRCh38
Transcripts
NM_001302643.1 NM_022082.3 XM_011528978.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4726605 c.1147G>T p.G383C Chr.20: 62966732-62966732 on Build GRCh38
109842 c.1139C>T p.A380V Chr.20: 62966724-62966724 on Build GRCh38
1413044 c.1134C>T p.A378A Chr.20: 62966719-62966719 on Build GRCh38
4448626 c.1117+1G>A p.? Chr.20: 62966581-62966581 on Build GRCh38
6160832 c.1133C>T p.A378V Chr.20: 62966718-62966718 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs62197279 Chr.20: 62966738-62966738 on Build GRCh38
rs918487092 Chr.20: 62966798-62966798 on Build GRCh38
rs375212323 Chr.20: 62966626-62966626 on Build GRCh38
rs374675240 Chr.20: 62966723-62966723 on Build GRCh38
rs368277499 Chr.20: 62966628-62966628 on Build GRCh38
rs775745540 Chr.20: 62966745-62966745 on Build GRCh38
rs45456594 Chr.20: 62966741-62966741 on Build GRCh38
rs779516672 Chr.20: 62966585-62966585 on Build GRCh38
rs5842409 Chr.20: 62966595-62966595 on Build GRCh38
rs752672533 Chr.20: 62966724-62966724 on Build GRCh38
rs776770149 Chr.20: 62966598-62966598 on Build GRCh38
rs200313034 Chr.20: 62966597-62966597 on Build GRCh38
rs999940052 Chr.20: 62966703-62966703 on Build GRCh38
rs146088564 Chr.20: 62966724-62966724 on Build GRCh38
rs772038305 Chr.20: 62966744-62966744 on Build GRCh38
rs553340506 Chr.20: 62966625-62966625 on Build GRCh38
rs371630761 Chr.20: 62966720-62966720 on Build GRCh38
rs201954865 Chr.20: 62966719-62966719 on Build GRCh38
rs116780417 Chr.20: 62966680-62966680 on Build GRCh38
rs565576713 Chr.20: 62966657-62966657 on Build GRCh38
rs200467016 Chr.20: 62966772-62966772 on Build GRCh38
rs985908065 Chr.20: 62966730-62966730 on Build GRCh38
rs777002593 Chr.20: 62966763-62966763 on Build GRCh38
rs397812887 Chr.20: 62966596-62966596 on Build GRCh38
rs535447189 Chr.20: 62966651-62966651 on Build GRCh38
rs760864056 Chr.20: 62966754-62966754 on Build GRCh38
rs757982227 Chr.20: 62966805-62966805 on Build GRCh38
rs367814195 Chr.20: 62966693-62966693 on Build GRCh38
rs141598974 Chr.20: 62966595-62966595 on Build GRCh38
rs757729486 Chr.20: 62966664-62966664 on Build GRCh38
rs779240177 Chr.20: 62966678-62966678 on Build GRCh38
rs767790015 Chr.20: 62966627-62966627 on Build GRCh38
rs764482505 Chr.20: 62966758-62966758 on Build GRCh38
rs35203370 Chr.20: 62966766-62966767 on Build GRCh38
rs111819136 Chr.20: 62966789-62966789 on Build GRCh38
rs746648856 Chr.20: 62966688-62966688 on Build GRCh38
rs779825324 Chr.20: 62966685-62966685 on Build GRCh38
rs750889263 Chr.20: 62966790-62966790 on Build GRCh38
rs771178867 Chr.20: 62966612-62966612 on Build GRCh38
rs575785128 Chr.20: 62966725-62966725 on Build GRCh38
rs1041348883 Chr.20: 62966682-62966682 on Build GRCh38
rs568449414 Chr.20: 62966632-62966632 on Build GRCh38
rs746806301 Chr.20: 62966586-62966586 on Build GRCh38
rs765781576 Chr.20: 62966808-62966808 on Build GRCh38
rs762107430 Chr.20: 62966768-62966768 on Build GRCh38
rs761824186 Chr.20: 62966610-62966610 on Build GRCh38
rs754515405 Chr.20: 62966735-62966735 on Build GRCh38
rs757781375 Chr.20: 62966684-62966684 on Build GRCh38
rs374528187 Chr.20: 62966660-62966660 on Build GRCh38
rs371579899 Chr.20: 62966645-62966645 on Build GRCh38

View Full Product Details