Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 178450203-178450476 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRKRA
Entrez Gene ID
8575
Gene Name
protein activator of interferon induced protein kinase EIF2AK2
Gene Aliases
DYT16, HSD14, PACT, RAX
UniGene
Hs.570274
Gene Chromosome Location
Chr.2: 178431414-178451240 on Build GRCh38
Transcripts
NM_001139517.1 NM_001139518.1 NM_001316362.1 NM_003690.4 XM_011512063.1 XM_011512066.1 XM_017005159.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6155413 c.52G>A p.A18T Chr.2: 178450392-178450392 on Build GRCh38
3757789 c.173A>G p.D58G Chr.2: 178450304-178450304 on Build GRCh38
6155415 c.72G>A p.P24P Chr.2: 178450372-178450372 on Build GRCh38
6155414 c.105G>A p.P35P Chr.2: 178450372-178450372 on Build GRCh38
6282143 c.182T>A p.V61D Chr.2: 178450262-178450262 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368860883 Chr.2: 178450450-178450450 on Build GRCh38
rs748210228 Chr.2: 178450453-178450453 on Build GRCh38
rs766343952 Chr.2: 178450369-178450369 on Build GRCh38
rs777378640 Chr.2: 178450411-178450411 on Build GRCh38
rs752176910 Chr.2: 178450398-178450398 on Build GRCh38
rs866097444 Chr.2: 178450415-178450415 on Build GRCh38
rs375307696 Chr.2: 178450421-178450421 on Build GRCh38
rs189979808 Chr.2: 178450244-178450244 on Build GRCh38
rs772470217 Chr.2: 178450291-178450291 on Build GRCh38
rs776733670 Chr.2: 178450447-178450447 on Build GRCh38
rs769066475 Chr.2: 178450438-178450438 on Build GRCh38
rs371966651 Chr.2: 178450359-178450359 on Build GRCh38
rs768718101 Chr.2: 178450316-178450316 on Build GRCh38
rs539630421 Chr.2: 178450274-178450274 on Build GRCh38
rs965736170 Chr.2: 178450443-178450443 on Build GRCh38
rs535247514 Chr.2: 178450427-178450427 on Build GRCh38
rs557598648 Chr.2: 178450433-178450433 on Build GRCh38
rs775966438 Chr.2: 178450297-178450297 on Build GRCh38
rs772423771 Chr.2: 178450430-178450430 on Build GRCh38
rs986066745 Chr.2: 178450381-178450381 on Build GRCh38
rs753131558 Chr.2: 178450231-178450231 on Build GRCh38
rs780703241 Chr.2: 178450397-178450397 on Build GRCh38
rs867139205 Chr.2: 178450243-178450244 on Build GRCh38
rs549677714 Chr.2: 178450363-178450363 on Build GRCh38
rs751447432 Chr.2: 178450372-178450372 on Build GRCh38
rs764047755 Chr.2: 178450413-178450414 on Build GRCh38
rs754883832 Chr.2: 178450378-178450378 on Build GRCh38
rs765855887 Chr.2: 178450415-178450416 on Build GRCh38
rs755656753 Chr.2: 178450400-178450400 on Build GRCh38
rs1005141587 Chr.2: 178450403-178450403 on Build GRCh38
rs200794293 Chr.2: 178450371-178450371 on Build GRCh38
rs776589927 Chr.2: 178450332-178450332 on Build GRCh38
rs779206948 Chr.2: 178450270-178450270 on Build GRCh38
rs62176111 Chr.2: 178450222-178450222 on Build GRCh38
rs750714380 Chr.2: 178450360-178450360 on Build GRCh38
rs762048114 Chr.2: 178450357-178450357 on Build GRCh38
rs61999302 Chr.2: 178450304-178450304 on Build GRCh38
rs911912217 Chr.2: 178450440-178450440 on Build GRCh38

View Full Product Details