Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 150150047-150150288 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLEKHO1
Entrez Gene ID
51177
Gene Name
pleckstrin homology domain containing O1
Gene Aliases
CKIP-1, CKIP1, JBP, OC120
UniGene
Hs.438824
Gene Chromosome Location
Chr.1: 150149434-150160065 on Build GRCh38
Transcripts
NM_001304722.1 NM_001304724.1 NM_016274.5 XM_017001420.1

SNP Detail

SNP ID Chromosome Location
rs782122632 Chr.1: 150150236-150150236 on Build GRCh38
rs587643363 Chr.1: 150150235-150150236 on Build GRCh38
rs189274971 Chr.1: 150150179-150150179 on Build GRCh38
rs781800628 Chr.1: 150150252-150150252 on Build GRCh38
rs782410256 Chr.1: 150150226-150150226 on Build GRCh38
rs782472584 Chr.1: 150150234-150150235 on Build GRCh38
rs76541931 Chr.1: 150150079-150150079 on Build GRCh38
rs76050098 Chr.1: 150150154-150150154 on Build GRCh38
rs782738833 Chr.1: 150150247-150150247 on Build GRCh38
rs782007643 Chr.1: 150150228-150150228 on Build GRCh38

View Full Product Details