Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 117157630-117157903 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC30A8
Entrez Gene ID
169026
Gene Name
solute carrier family 30 member 8
Gene Aliases
ZNT8, ZnT-8
UniGene
Hs.532270
Gene Chromosome Location
Chr.8: 116950273-117176714 on Build GRCh38
Transcripts
NM_001172811.1 NM_001172813.1 NM_001172814.1 NM_001172815.2 NM_173851.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
273263 c.463A>T p.T155S Chr.8: 117157735-117157735 on Build GRCh38
1226271 c.516G>T p.Q172H Chr.8: 117157788-117157788 on Build GRCh38
3644523 c.438C>T p.L146L Chr.8: 117157710-117157710 on Build GRCh38
3929350 c.553G>A p.A185T Chr.8: 117157825-117157825 on Build GRCh38
1180401 c.432C>T p.A144A Chr.8: 117157704-117157704 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1042096683 Chr.8: 117157865-117157865 on Build GRCh38
rs199894401 Chr.8: 117157659-117157659 on Build GRCh38
rs760270538 Chr.8: 117157690-117157690 on Build GRCh38
rs199875716 Chr.8: 117157834-117157834 on Build GRCh38
rs764048971 Chr.8: 117157738-117157738 on Build GRCh38
rs1020396629 Chr.8: 117157661-117157661 on Build GRCh38
rs1018852833 Chr.8: 117157719-117157719 on Build GRCh38
rs755241418 Chr.8: 117157833-117157833 on Build GRCh38
rs769119495 Chr.8: 117157862-117157862 on Build GRCh38
rs751432460 Chr.8: 117157739-117157739 on Build GRCh38
rs747531216 Chr.8: 117157697-117157697 on Build GRCh38
rs745417013 Chr.8: 117157845-117157845 on Build GRCh38
rs769312132 Chr.8: 117157669-117157669 on Build GRCh38
rs757129691 Chr.8: 117157740-117157740 on Build GRCh38
rs764763610 Chr.8: 117157708-117157708 on Build GRCh38
rs377289568 Chr.8: 117157877-117157877 on Build GRCh38
rs946052598 Chr.8: 117157860-117157860 on Build GRCh38
rs578012694 Chr.8: 117157748-117157748 on Build GRCh38
rs748727258 Chr.8: 117157796-117157796 on Build GRCh38
rs374026995 Chr.8: 117157867-117157867 on Build GRCh38
rs779581100 Chr.8: 117157792-117157792 on Build GRCh38
rs765852756 Chr.8: 117157694-117157694 on Build GRCh38
rs980668978 Chr.8: 117157804-117157804 on Build GRCh38
rs146835671 Chr.8: 117157741-117157741 on Build GRCh38
rs778095166 Chr.8: 117157844-117157844 on Build GRCh38
rs373292003 Chr.8: 117157665-117157665 on Build GRCh38
rs142407509 Chr.8: 117157726-117157726 on Build GRCh38
rs762742030 Chr.8: 117157808-117157808 on Build GRCh38
rs141202988 Chr.8: 117157832-117157832 on Build GRCh38
rs201797531 Chr.8: 117157802-117157802 on Build GRCh38
rs73317647 Chr.8: 117157765-117157765 on Build GRCh38
rs774963524 Chr.8: 117157670-117157670 on Build GRCh38
rs751350884 Chr.8: 117157816-117157816 on Build GRCh38
rs756406161 Chr.8: 117157758-117157758 on Build GRCh38
rs181713403 Chr.8: 117157824-117157824 on Build GRCh38
rs758949776 Chr.8: 117157800-117157800 on Build GRCh38
rs759297376 Chr.8: 117157707-117157707 on Build GRCh38
rs146555947 Chr.8: 117157812-117157812 on Build GRCh38
rs995417294 Chr.8: 117157875-117157875 on Build GRCh38
rs767594541 Chr.8: 117157825-117157825 on Build GRCh38
rs775084835 Chr.8: 117157806-117157806 on Build GRCh38
rs867137285 Chr.8: 117157818-117157818 on Build GRCh38
rs370682947 Chr.8: 117157840-117157840 on Build GRCh38
rs780088312 Chr.8: 117157760-117157760 on Build GRCh38
rs201697165 Chr.8: 117157817-117157817 on Build GRCh38
rs776248662 Chr.8: 117157797-117157797 on Build GRCh38
rs376458891 Chr.8: 117157710-117157710 on Build GRCh38
rs145677283 Chr.8: 117157766-117157766 on Build GRCh38
rs761849730 Chr.8: 117157823-117157823 on Build GRCh38
rs560531972 Chr.8: 117157773-117157773 on Build GRCh38
rs185043998 Chr.8: 117157841-117157841 on Build GRCh38
rs190337127 Chr.8: 117157666-117157666 on Build GRCh38
rs775987808 Chr.8: 117157696-117157696 on Build GRCh38

View Full Product Details