Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 46267901-46268150 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TTC38
Entrez Gene ID
55020
Gene Name
tetratricopeptide repeat domain 38
Gene Aliases
LL22NC03-5H6.5
UniGene
Hs.435832
Gene Chromosome Location
Chr.22: 46267964-46294008 on Build GRCh38
Transcripts
NM_017931.2 XM_011530259.1 XM_011530260.2

SNP Detail

SNP ID Chromosome Location
rs764477282 Chr.22: 46268019-46268019 on Build GRCh38
rs774851108 Chr.22: 46268006-46268006 on Build GRCh38
rs954280714 Chr.22: 46268044-46268044 on Build GRCh38
rs1013253363 Chr.22: 46268001-46268001 on Build GRCh38
rs758531013 Chr.22: 46268038-46268038 on Build GRCh38
rs946809468 Chr.22: 46267969-46267969 on Build GRCh38
rs563970921 Chr.22: 46268053-46268053 on Build GRCh38
rs961250459 Chr.22: 46268064-46268064 on Build GRCh38
rs775146670 Chr.22: 46268015-46268015 on Build GRCh38
rs759161157 Chr.22: 46268004-46268004 on Build GRCh38
rs999865064 Chr.22: 46267965-46267965 on Build GRCh38
rs556059445 Chr.22: 46268018-46268018 on Build GRCh38
rs745362525 Chr.22: 46268121-46268121 on Build GRCh38
rs767278372 Chr.22: 46268014-46268014 on Build GRCh38
rs1033125153 Chr.22: 46267971-46267971 on Build GRCh38
rs866503246 Chr.22: 46268124-46268124 on Build GRCh38
rs867259037 Chr.22: 46267979-46267979 on Build GRCh38
rs935757064 Chr.22: 46268123-46268123 on Build GRCh38
rs112960550 Chr.22: 46267990-46267990 on Build GRCh38
rs747188517 Chr.22: 46268045-46268045 on Build GRCh38
rs765771865 Chr.22: 46268023-46268023 on Build GRCh38
rs770964175 Chr.22: 46268103-46268103 on Build GRCh38
rs749147411 Chr.22: 46268066-46268066 on Build GRCh38
rs924379673 Chr.22: 46268077-46268077 on Build GRCh38
rs572681723 Chr.22: 46268020-46268020 on Build GRCh38
rs902961190 Chr.22: 46267959-46267959 on Build GRCh38
rs762324935 Chr.22: 46268021-46268021 on Build GRCh38
rs774501552 Chr.22: 46268120-46268120 on Build GRCh38
rs921269455 Chr.22: 46268041-46268041 on Build GRCh38
rs974091141 Chr.22: 46268036-46268036 on Build GRCh38
rs990969464 Chr.22: 46268089-46268089 on Build GRCh38
rs750496594 Chr.22: 46268025-46268025 on Build GRCh38
rs773980399 Chr.22: 46267998-46267998 on Build GRCh38
rs1054088609 Chr.22: 46268125-46268125 on Build GRCh38
rs960874798 Chr.22: 46267947-46267951 on Build GRCh38
rs1041477140 Chr.22: 46267952-46267952 on Build GRCh38
rs533037119 Chr.22: 46268055-46268055 on Build GRCh38
rs757737391 Chr.22: 46268022-46268022 on Build GRCh38
rs540998469 Chr.22: 46268040-46268040 on Build GRCh38
rs770682248 Chr.22: 46268049-46268049 on Build GRCh38
rs924497619 Chr.22: 46267957-46267972 on Build GRCh38

View Full Product Details