Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 53299843-53300072 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C12orf10
Entrez Gene ID
60314
Gene Name
chromosome 12 open reading frame 10
Gene Aliases
Gamm1, MST024, MSTP024, MYG, MYG1
UniGene
Hs.655988
Gene Chromosome Location
Chr.12: 53299686-53307181 on Build GRCh38
Transcripts
NM_021640.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4666688 c.109T>C p.S37P Chr.12: 53299846-53299846 on Build GRCh38
7426225 c.194T>C p.L65P Chr.12: 53299931-53299931 on Build GRCh38
6242802 c.171C>T p.D57D Chr.12: 53299908-53299908 on Build GRCh38
7119990 c.162C>G p.F54L Chr.12: 53299899-53299899 on Build GRCh38
5964570 c.154G>T p.G52C Chr.12: 53299891-53299891 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761408410 Chr.12: 53300005-53300005 on Build GRCh38
rs143971152 Chr.12: 53299880-53299880 on Build GRCh38
rs200943581 Chr.12: 53299943-53299943 on Build GRCh38
rs550128329 Chr.12: 53299930-53299930 on Build GRCh38
rs547345622 Chr.12: 53299912-53299912 on Build GRCh38
rs767213154 Chr.12: 53299950-53299950 on Build GRCh38
rs143220786 Chr.12: 53299921-53299921 on Build GRCh38
rs775600170 Chr.12: 53299941-53299941 on Build GRCh38
rs753249019 Chr.12: 53299952-53299952 on Build GRCh38
rs762575983 Chr.12: 53299878-53299878 on Build GRCh38
rs148637045 Chr.12: 53299900-53299900 on Build GRCh38
rs1004415213 Chr.12: 53299884-53299884 on Build GRCh38
rs770180591 Chr.12: 53299963-53299963 on Build GRCh38
rs769753014 Chr.12: 53299973-53299973 on Build GRCh38
rs993809420 Chr.12: 53299862-53299862 on Build GRCh38
rs769806164 Chr.12: 53299937-53299937 on Build GRCh38
rs766718398 Chr.12: 53299899-53299899 on Build GRCh38
rs777785462 Chr.12: 53299954-53299954 on Build GRCh38
rs779411421 Chr.12: 53299908-53299908 on Build GRCh38
rs758461666 Chr.12: 53299953-53299953 on Build GRCh38
rs970794751 Chr.12: 53299896-53299896 on Build GRCh38
rs777947031 Chr.12: 53299927-53299927 on Build GRCh38
rs754098913 Chr.12: 53299951-53299951 on Build GRCh38
rs773676479 Chr.12: 53299998-53299998 on Build GRCh38
rs756486053 Chr.12: 53299890-53299890 on Build GRCh38
rs1015840185 Chr.12: 53299888-53299888 on Build GRCh38
rs539064889 Chr.12: 53299958-53299958 on Build GRCh38
rs536309702 Chr.12: 53300053-53300053 on Build GRCh38
rs138701588 Chr.12: 53299993-53299993 on Build GRCh38
rs936596953 Chr.12: 53300023-53300023 on Build GRCh38
rs770759149 Chr.12: 53299933-53299933 on Build GRCh38
rs749302522 Chr.12: 53299992-53299992 on Build GRCh38
rs765274304 Chr.12: 53299863-53299863 on Build GRCh38
rs375146043 Chr.12: 53299986-53299986 on Build GRCh38
rs745619554 Chr.12: 53299970-53299970 on Build GRCh38
rs755420021 Chr.12: 53299901-53299901 on Build GRCh38
rs567432764 Chr.12: 53299867-53299867 on Build GRCh38
rs763748665 Chr.12: 53299945-53299945 on Build GRCh38
rs1026675621 Chr.12: 53299889-53299889 on Build GRCh38
rs781744738 Chr.12: 53299911-53299911 on Build GRCh38
rs760128840 Chr.12: 53299935-53299935 on Build GRCh38
rs888624035 Chr.12: 53299879-53299879 on Build GRCh38
rs572508056 Chr.12: 53300013-53300013 on Build GRCh38
rs369388636 Chr.12: 53299990-53299990 on Build GRCh38
rs781499872 Chr.12: 53299966-53299966 on Build GRCh38
rs777161707 Chr.12: 53299904-53299904 on Build GRCh38
rs761304645 Chr.12: 53299947-53299947 on Build GRCh38

View Full Product Details