Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 35941130-35941393 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CFAP47
Entrez Gene ID
286464
Gene Name
cilia and flagella associated protein 47
Gene Aliases
CHDC2, CXorf22, CXorf30, CXorf59
UniGene
Hs.376425
Gene Chromosome Location
Chr.X: 35919734-36385319 on Build GRCh38
Transcripts
NM_001304548.1 NM_152632.3 XM_017029452.1 XM_017029453.1

SNP Detail

SNP ID Chromosome Location
rs148706541 Chr.X: 35941295-35941295 on Build GRCh38
rs142281214 Chr.X: 35941364-35941364 on Build GRCh38
rs758440622 Chr.X: 35941348-35941348 on Build GRCh38
rs772162892 Chr.X: 35941320-35941320 on Build GRCh38
rs764247271 Chr.X: 35941250-35941250 on Build GRCh38
rs868489188 Chr.X: 35941294-35941294 on Build GRCh38
rs1024088621 Chr.X: 35941198-35941198 on Build GRCh38
rs750947586 Chr.X: 35941330-35941330 on Build GRCh38
rs922498095 Chr.X: 35941186-35941186 on Build GRCh38
rs370681633 Chr.X: 35941354-35941354 on Build GRCh38
rs751555600 Chr.X: 35941213-35941213 on Build GRCh38
rs933685147 Chr.X: 35941229-35941229 on Build GRCh38
rs373042953 Chr.X: 35941359-35941359 on Build GRCh38
rs754194229 Chr.X: 35941296-35941296 on Build GRCh38
rs754037020 Chr.X: 35941279-35941279 on Build GRCh38
rs748300791 Chr.X: 35941252-35941252 on Build GRCh38
rs773507455 Chr.X: 35941347-35941347 on Build GRCh38
rs770378384 Chr.X: 35941262-35941262 on Build GRCh38
rs780015849 Chr.X: 35941349-35941349 on Build GRCh38
rs946417076 Chr.X: 35941265-35941265 on Build GRCh38
rs775757114 Chr.X: 35941158-35941159 on Build GRCh38
rs376295810 Chr.X: 35941240-35941240 on Build GRCh38
rs978310879 Chr.X: 35941230-35941230 on Build GRCh38
rs760771714 Chr.X: 35941161-35941161 on Build GRCh38
rs762203457 Chr.X: 35941316-35941316 on Build GRCh38
rs957282493 Chr.X: 35941263-35941263 on Build GRCh38

View Full Product Details