Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 103414851-103415102 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCAF13
Entrez Gene ID
25879
Gene Name
DDB1 and CUL4 associated factor 13
Gene Aliases
GM83, HSPC064, WDSOF1
UniGene
Hs.532265
Gene Chromosome Location
Chr.8: 103414714-103443453 on Build GRCh38
Transcripts
NM_015420.6 NM_030780.4 NR_102337.1 NR_102338.1 XM_017013877.1

Gene Symbol
SLC25A32
Entrez Gene ID
81034
Gene Name
solute carrier family 25 member 32
Gene Aliases
MFT, MFTC, RREI
UniGene
Hs.532265
Gene Chromosome Location
Chr.8: 103398638-103415335 on Build GRCh38
Transcripts
NM_015420.6 NM_030780.4 NR_102337.1 NR_102338.1 XM_017013877.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6193758 c.70G>C p.E24Q Chr.8: 103414868-103414868 on Build GRCh38
4841806 c.42G>A p.W14* Chr.8: 103414896-103414896 on Build GRCh38
1226125 c.8G>C p.G3A Chr.8: 103414930-103414930 on Build GRCh38
1453734 c.83C>T p.A28V Chr.8: 103414855-103414855 on Build GRCh38
6431158 c.56G>T p.R19L Chr.8: 103414882-103414882 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs897683832 Chr.8: 103415075-103415075 on Build GRCh38
rs766544817 Chr.8: 103414919-103414920 on Build GRCh38
rs779935311 Chr.8: 103415039-103415039 on Build GRCh38
rs928387933 Chr.8: 103414978-103414978 on Build GRCh38
rs182570628 Chr.8: 103415076-103415076 on Build GRCh38
rs765910763 Chr.8: 103414903-103414903 on Build GRCh38
rs764280139 Chr.8: 103415077-103415077 on Build GRCh38
rs760825140 Chr.8: 103415016-103415016 on Build GRCh38
rs148406806 Chr.8: 103415062-103415062 on Build GRCh38
rs745653877 Chr.8: 103414951-103414951 on Build GRCh38
rs766656853 Chr.8: 103414923-103414923 on Build GRCh38
rs374257876 Chr.8: 103415066-103415066 on Build GRCh38
rs763459527 Chr.8: 103414912-103414912 on Build GRCh38
rs1012703713 Chr.8: 103415040-103415040 on Build GRCh38
rs773839769 Chr.8: 103414911-103414911 on Build GRCh38
rs750253444 Chr.8: 103415036-103415036 on Build GRCh38
rs778880688 Chr.8: 103415033-103415033 on Build GRCh38
rs752932122 Chr.8: 103415015-103415015 on Build GRCh38
rs199877106 Chr.8: 103415028-103415028 on Build GRCh38
rs757182453 Chr.8: 103414949-103414949 on Build GRCh38
rs771664298 Chr.8: 103414963-103414963 on Build GRCh38
rs543197401 Chr.8: 103415037-103415037 on Build GRCh38
rs771707465 Chr.8: 103414877-103414877 on Build GRCh38
rs780867938 Chr.8: 103415056-103415056 on Build GRCh38
rs775796602 Chr.8: 103415070-103415070 on Build GRCh38
rs767788695 Chr.8: 103415008-103415008 on Build GRCh38
rs377166611 Chr.8: 103414992-103414992 on Build GRCh38
rs749232294 Chr.8: 103414945-103414945 on Build GRCh38
rs776213749 Chr.8: 103414987-103414987 on Build GRCh38
rs747668941 Chr.8: 103415059-103415059 on Build GRCh38
rs1025803042 Chr.8: 103414982-103414982 on Build GRCh38
rs756176285 Chr.8: 103414941-103414941 on Build GRCh38
rs767072080 Chr.8: 103415025-103415025 on Build GRCh38
rs941958180 Chr.8: 103415052-103415052 on Build GRCh38
rs990243771 Chr.8: 103414946-103414946 on Build GRCh38
rs777774840 Chr.8: 103414943-103414943 on Build GRCh38
rs971513571 Chr.8: 103415061-103415061 on Build GRCh38
rs768292692 Chr.8: 103414981-103414981 on Build GRCh38
rs760233515 Chr.8: 103414892-103414892 on Build GRCh38
rs761950146 Chr.8: 103415082-103415082 on Build GRCh38
rs746663173 Chr.8: 103415042-103415042 on Build GRCh38
rs908674888 Chr.8: 103415066-103415067 on Build GRCh38
rs377666609 Chr.8: 103414937-103414937 on Build GRCh38
rs768382507 Chr.8: 103415054-103415054 on Build GRCh38
rs372902714 Chr.8: 103414965-103414965 on Build GRCh38
rs3814106 Chr.8: 103415006-103415006 on Build GRCh38
rs977227616 Chr.8: 103414974-103414974 on Build GRCh38
rs143980576 Chr.8: 103415067-103415067 on Build GRCh38
rs763398540 Chr.8: 103414989-103414989 on Build GRCh38
rs776685812 Chr.8: 103415074-103415074 on Build GRCh38
rs778595960 Chr.8: 103414950-103414950 on Build GRCh38
rs991567038 Chr.8: 103415022-103415022 on Build GRCh38
rs775190762 Chr.8: 103414891-103414891 on Build GRCh38
rs1032698210 Chr.8: 103415002-103415002 on Build GRCh38
rs370878481 Chr.8: 103414940-103414940 on Build GRCh38
rs753905788 Chr.8: 103415078-103415078 on Build GRCh38
rs746574905 Chr.8: 103414971-103414971 on Build GRCh38
rs555069499 Chr.8: 103415060-103415060 on Build GRCh38
rs755173340 Chr.8: 103414927-103414927 on Build GRCh38
rs757191177 Chr.8: 103415031-103415031 on Build GRCh38
rs751908054 Chr.8: 103414925-103414925 on Build GRCh38
rs370320899 Chr.8: 103415000-103415000 on Build GRCh38
rs1049823457 Chr.8: 103414875-103414875 on Build GRCh38

View Full Product Details