Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 53425804-53426029 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMHR2
Entrez Gene ID
269
Gene Name
anti-Mullerian hormone receptor type 2
Gene Aliases
AMHR, MISR2, MISRII, MRII
UniGene
Hs.659889
Gene Chromosome Location
Chr.12: 53423855-53431672 on Build GRCh38
Transcripts
NM_001164690.1 NM_001164691.1 NM_020547.2 XM_011538173.1 XM_011538174.1 XM_011538176.1 XM_011538178.1 XM_011538179.1 XM_011538180.1 XM_011538181.1 XM_011538183.2 XM_011538184.2 XM_011538185.1 XM_011538186.2 XM_011538187.2 XM_017019179.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5375474 c.748C>T p.P250S Chr.12: 53425815-53425815 on Build GRCh38
5069583 c.793_794insG p.P268fs*30 Chr.12: 53425860-53425861 on Build GRCh38
5375476 c.794G>A p.R265Q Chr.12: 53425861-53425861 on Build GRCh38
5375475 c.749C>T p.P250L Chr.12: 53425816-53425816 on Build GRCh38
1677022 c.818C>T p.S273F Chr.12: 53425885-53425885 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772542750 Chr.12: 53425880-53425880 on Build GRCh38
rs555747490 Chr.12: 53425981-53425981 on Build GRCh38
rs752826964 Chr.12: 53425926-53425926 on Build GRCh38
rs774742716 Chr.12: 53425952-53425953 on Build GRCh38
rs369599312 Chr.12: 53425860-53425860 on Build GRCh38
rs188490937 Chr.12: 53425974-53425974 on Build GRCh38
rs748586967 Chr.12: 53425875-53425875 on Build GRCh38
rs746905091 Chr.12: 53425842-53425842 on Build GRCh38
rs779570019 Chr.12: 53425867-53425867 on Build GRCh38
rs373621190 Chr.12: 53425843-53425843 on Build GRCh38
rs1027570794 Chr.12: 53425984-53425984 on Build GRCh38
rs757132217 Chr.12: 53425970-53425970 on Build GRCh38
rs539695176 Chr.12: 53425913-53425913 on Build GRCh38
rs115267458 Chr.12: 53425829-53425829 on Build GRCh38
rs762781092 Chr.12: 53425939-53425939 on Build GRCh38
rs758179091 Chr.12: 53425832-53425832 on Build GRCh38
rs761223361 Chr.12: 53425892-53425892 on Build GRCh38
rs569194253 Chr.12: 53425879-53425879 on Build GRCh38
rs773878247 Chr.12: 53425891-53425891 on Build GRCh38
rs777898078 Chr.12: 53425864-53425868 on Build GRCh38
rs748387033 Chr.12: 53425860-53425861 on Build GRCh38
rs765559673 Chr.12: 53425916-53425916 on Build GRCh38
rs776713395 Chr.12: 53425905-53425905 on Build GRCh38
rs780980821 Chr.12: 53425971-53425971 on Build GRCh38
rs745524273 Chr.12: 53425864-53425864 on Build GRCh38
rs781413894 Chr.12: 53425852-53425855 on Build GRCh38
rs769854694 Chr.12: 53425861-53425861 on Build GRCh38
rs751506278 Chr.12: 53425964-53425964 on Build GRCh38
rs1016766065 Chr.12: 53425878-53425878 on Build GRCh38
rs138308620 Chr.12: 53425902-53425902 on Build GRCh38
rs374370282 Chr.12: 53425861-53425861 on Build GRCh38
rs769550210 Chr.12: 53425866-53425866 on Build GRCh38
rs991274756 Chr.12: 53425947-53425947 on Build GRCh38
rs774776142 Chr.12: 53425836-53425836 on Build GRCh38
rs771519500 Chr.12: 53425940-53425941 on Build GRCh38
rs377740685 Chr.12: 53425956-53425956 on Build GRCh38
rs114082777 Chr.12: 53425898-53425898 on Build GRCh38
rs746203805 Chr.12: 53425955-53425956 on Build GRCh38
rs139495947 Chr.12: 53425953-53425954 on Build GRCh38
rs917014741 Chr.12: 53425947-53425947 on Build GRCh38

View Full Product Details