Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 148845473-148845734 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPB1
Entrez Gene ID
1360
Gene Name
carboxypeptidase B1
Gene Aliases
CPB, PASP, PCPB
UniGene
Hs.477891
Gene Chromosome Location
Chr.3: 148827801-148860187 on Build GRCh38
Transcripts
NM_001871.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6269207 c.859G>T p.A287S Chr.3: 148845504-148845504 on Build GRCh38
4982410 c.872G>A p.R291H Chr.3: 148845517-148845517 on Build GRCh38
160172 c.961G>A p.G321S Chr.3: 148845606-148845606 on Build GRCh38
5596531 c.966G>A p.E322E Chr.3: 148845611-148845611 on Build GRCh38
1039668 c.882C>A p.L294L Chr.3: 148845527-148845527 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778982431 Chr.3: 148845517-148845517 on Build GRCh38
rs151225556 Chr.3: 148845605-148845605 on Build GRCh38
rs780284562 Chr.3: 148845649-148845649 on Build GRCh38
rs756976312 Chr.3: 148845606-148845606 on Build GRCh38
rs776970460 Chr.3: 148845664-148845664 on Build GRCh38
rs966121951 Chr.3: 148845613-148845613 on Build GRCh38
rs914844293 Chr.3: 148845683-148845683 on Build GRCh38
rs376118016 Chr.3: 148845652-148845652 on Build GRCh38
rs372618980 Chr.3: 148845595-148845595 on Build GRCh38
rs777859924 Chr.3: 148845500-148845500 on Build GRCh38
rs941936702 Chr.3: 148845641-148845641 on Build GRCh38
rs866231841 Chr.3: 148845628-148845628 on Build GRCh38
rs752059114 Chr.3: 148845535-148845535 on Build GRCh38
rs778837863 Chr.3: 148845642-148845642 on Build GRCh38
rs762448526 Chr.3: 148845666-148845666 on Build GRCh38
rs1056872917 Chr.3: 148845687-148845688 on Build GRCh38
rs866048233 Chr.3: 148845572-148845572 on Build GRCh38
rs201123161 Chr.3: 148845557-148845557 on Build GRCh38
rs767292387 Chr.3: 148845587-148845587 on Build GRCh38
rs772211550 Chr.3: 148845647-148845647 on Build GRCh38
rs748085674 Chr.3: 148845497-148845497 on Build GRCh38
rs1001634903 Chr.3: 148845678-148845678 on Build GRCh38
rs748997009 Chr.3: 148845504-148845504 on Build GRCh38
rs572500402 Chr.3: 148845650-148845650 on Build GRCh38
rs1047636855 Chr.3: 148845545-148845545 on Build GRCh38
rs770785642 Chr.3: 148845506-148845506 on Build GRCh38
rs562228830 Chr.3: 148845569-148845569 on Build GRCh38
rs948361279 Chr.3: 148845705-148845705 on Build GRCh38
rs777526007 Chr.3: 148845603-148845603 on Build GRCh38
rs746146300 Chr.3: 148845521-148845521 on Build GRCh38
rs756057467 Chr.3: 148845499-148845499 on Build GRCh38
rs762060736 Chr.3: 148845560-148845560 on Build GRCh38
rs199791381 Chr.3: 148845546-148845546 on Build GRCh38
rs751166132 Chr.3: 148845576-148845576 on Build GRCh38
rs151168626 Chr.3: 148845673-148845673 on Build GRCh38
rs759321381 Chr.3: 148845583-148845583 on Build GRCh38
rs752190410 Chr.3: 148845592-148845592 on Build GRCh38
rs139558909 Chr.3: 148845688-148845688 on Build GRCh38
rs142211299 Chr.3: 148845516-148845516 on Build GRCh38
rs533425814 Chr.3: 148845704-148845704 on Build GRCh38
rs3765131 Chr.3: 148845706-148845706 on Build GRCh38
rs745659583 Chr.3: 148845646-148845646 on Build GRCh38
rs976655134 Chr.3: 148845622-148845622 on Build GRCh38
rs775486211 Chr.3: 148845532-148845532 on Build GRCh38
rs564586318 Chr.3: 148845674-148845674 on Build GRCh38
rs772138022 Chr.3: 148845527-148845527 on Build GRCh38

View Full Product Details