Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 109696343-109696568 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CASP6
Entrez Gene ID
839
Gene Name
caspase 6
Gene Aliases
MCH2
UniGene
Hs.654616
Gene Chromosome Location
Chr.4: 109688629-109703473 on Build GRCh38
Transcripts
NM_001226.3 NM_032992.2 NR_133012.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4839279 c.296A>C p.K99T Chr.4: 109696421-109696421 on Build GRCh38
1049966 c.292C>T p.L98F Chr.4: 109696425-109696425 on Build GRCh38
6620599 c.238G>T p.D80Y Chr.4: 109696479-109696479 on Build GRCh38
1618257 c.237A>T p.S79S Chr.4: 109696480-109696480 on Build GRCh38
1618256 c.295A>G p.K99E Chr.4: 109696422-109696422 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs914629289 Chr.4: 109696514-109696514 on Build GRCh38
rs759168093 Chr.4: 109696411-109696411 on Build GRCh38
rs543685141 Chr.4: 109696431-109696431 on Build GRCh38
rs774000583 Chr.4: 109696403-109696403 on Build GRCh38
rs956225546 Chr.4: 109696492-109696492 on Build GRCh38
rs750484000 Chr.4: 109696488-109696488 on Build GRCh38
rs377529968 Chr.4: 109696421-109696421 on Build GRCh38
rs373103632 Chr.4: 109696512-109696512 on Build GRCh38
rs766749828 Chr.4: 109696432-109696434 on Build GRCh38
rs781774694 Chr.4: 109696375-109696375 on Build GRCh38
rs760424006 Chr.4: 109696415-109696415 on Build GRCh38
rs375725324 Chr.4: 109696535-109696535 on Build GRCh38
rs574926307 Chr.4: 109696414-109696414 on Build GRCh38
rs373891737 Chr.4: 109696408-109696408 on Build GRCh38
rs998292877 Chr.4: 109696405-109696405 on Build GRCh38
rs148210591 Chr.4: 109696413-109696413 on Build GRCh38
rs780172118 Chr.4: 109696502-109696502 on Build GRCh38
rs761880056 Chr.4: 109696467-109696467 on Build GRCh38
rs753833596 Chr.4: 109696432-109696432 on Build GRCh38
rs201166197 Chr.4: 109696394-109696394 on Build GRCh38
rs764001587 Chr.4: 109696430-109696430 on Build GRCh38
rs748792961 Chr.4: 109696382-109696382 on Build GRCh38
rs5030561 Chr.4: 109696493-109696493 on Build GRCh38
rs187930015 Chr.4: 109696481-109696481 on Build GRCh38

View Full Product Details