Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 174571786-174572025 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WIPF1
Entrez Gene ID
7456
Gene Name
WAS/WASL interacting protein family member 1
Gene Aliases
PRPL-2, WAS2, WASPIP, WIP
UniGene
Hs.128067
Gene Chromosome Location
Chr.2: 174559574-174682899 on Build GRCh38
Transcripts
NM_001077269.1 NM_003387.4 XM_011511780.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4864952 c.842C>T p.A281V Chr.2: 174571963-174571963 on Build GRCh38
1009777 c.842C>T p.A281V Chr.2: 174571963-174571963 on Build GRCh38
1009775 c.866A>G p.N289S Chr.2: 174571939-174571939 on Build GRCh38
3391133 c.981C>T p.G327G Chr.2: 174571824-174571824 on Build GRCh38
1582491 c.988G>A p.E330K Chr.2: 174571817-174571817 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776988354 Chr.2: 174571899-174571899 on Build GRCh38
rs764421341 Chr.2: 174571813-174571813 on Build GRCh38
rs369696922 Chr.2: 174571962-174571962 on Build GRCh38
rs909498834 Chr.2: 174571876-174571876 on Build GRCh38
rs762111512 Chr.2: 174571902-174571902 on Build GRCh38
rs769149570 Chr.2: 174571943-174571945 on Build GRCh38
rs369153153 Chr.2: 174571976-174571976 on Build GRCh38
rs557450984 Chr.2: 174571911-174571911 on Build GRCh38
rs575913592 Chr.2: 174571912-174571912 on Build GRCh38
rs369736015 Chr.2: 174571957-174571957 on Build GRCh38
rs745430381 Chr.2: 174571867-174571878 on Build GRCh38
rs770409036 Chr.2: 174571822-174571822 on Build GRCh38
rs745551139 Chr.2: 174571896-174571896 on Build GRCh38
rs779235427 Chr.2: 174571948-174571948 on Build GRCh38
rs754336200 Chr.2: 174571925-174571925 on Build GRCh38
rs201234000 Chr.2: 174571818-174571818 on Build GRCh38
rs546654911 Chr.2: 174571817-174571817 on Build GRCh38
rs948386388 Chr.2: 174571895-174571895 on Build GRCh38
rs891642872 Chr.2: 174571945-174571945 on Build GRCh38
rs780660555 Chr.2: 174571879-174571879 on Build GRCh38
rs746027265 Chr.2: 174571865-174571865 on Build GRCh38
rs923133424 Chr.2: 174571882-174571882 on Build GRCh38
rs112447307 Chr.2: 174571848-174571848 on Build GRCh38
rs1040586347 Chr.2: 174571869-174571869 on Build GRCh38
rs146135868 Chr.2: 174571916-174571916 on Build GRCh38
rs751419161 Chr.2: 174571830-174571830 on Build GRCh38
rs779265386 Chr.2: 174571857-174571857 on Build GRCh38
rs764189324 Chr.2: 174571875-174571875 on Build GRCh38
rs929519056 Chr.2: 174571864-174571864 on Build GRCh38
rs764807602 Chr.2: 174571821-174571821 on Build GRCh38
rs375093864 Chr.2: 174571931-174571931 on Build GRCh38
rs1025528020 Chr.2: 174571985-174571985 on Build GRCh38
rs943312416 Chr.2: 174571851-174571851 on Build GRCh38
rs772015838 Chr.2: 174571868-174571868 on Build GRCh38
rs769490496 Chr.2: 174571903-174571903 on Build GRCh38
rs781658089 Chr.2: 174571978-174571978 on Build GRCh38
rs887567339 Chr.2: 174571920-174571920 on Build GRCh38
rs755969493 Chr.2: 174571946-174571946 on Build GRCh38
rs779500955 Chr.2: 174571935-174571935 on Build GRCh38
rs772963293 Chr.2: 174571990-174571990 on Build GRCh38
rs757927442 Chr.2: 174571933-174571935 on Build GRCh38
rs1052917202 Chr.2: 174571941-174571941 on Build GRCh38
rs781155820 Chr.2: 174571837-174571837 on Build GRCh38
rs754868175 Chr.2: 174571835-174571835 on Build GRCh38
rs201094141 Chr.2: 174571964-174571964 on Build GRCh38
rs556678311 Chr.2: 174571942-174571943 on Build GRCh38
rs752552539 Chr.2: 174571932-174571932 on Build GRCh38
rs147080148 Chr.2: 174571827-174571827 on Build GRCh38
rs757556263 Chr.2: 174571856-174571856 on Build GRCh38
rs770110301 Chr.2: 174571986-174571986 on Build GRCh38
rs199839276 Chr.2: 174571918-174571918 on Build GRCh38
rs762533332 Chr.2: 174571995-174571995 on Build GRCh38
rs747496276 Chr.2: 174571893-174571893 on Build GRCh38

View Full Product Details