Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 137252859-137253084 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C9orf173-AS1
Entrez Gene ID
100129722
Gene Name
C9orf173 antisense RNA 1
Gene Aliases
-
UniGene
Hs.656347
Gene Chromosome Location
Chr.9: 137250219-137253497 on Build GRCh38
Transcripts
NR_038389.1 NM_001004353.3 NM_001256699.1 NM_001256700.1 NM_001256701.1 NR_046339.1 NR_046340.1 XM_006717113.2 XM_006717115.2

Gene Symbol
STPG3
Entrez Gene ID
441476
Gene Name
sperm-tail PG-rich repeat containing 3
Gene Aliases
C9orf173
UniGene
Hs.372640
Gene Chromosome Location
Chr.9: 137250840-137253483 on Build GRCh38
Transcripts
NR_038389.1 NM_001004353.3 NM_001256699.1 NM_001256700.1 NM_001256701.1 NR_046339.1 NR_046340.1 XM_006717113.2 XM_006717115.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5777084 c.739C>T p.R247C Chr.9: 137252908-137252908 on Build GRCh38
6501200 c.746C>T p.P249L Chr.9: 137252915-137252915 on Build GRCh38
4651675 c.740G>A p.R247H Chr.9: 137252909-137252909 on Build GRCh38
4621371 c.712C>G p.L238V Chr.9: 137252881-137252881 on Build GRCh38
4939030 c.744G>A p.S248S Chr.9: 137252913-137252913 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1028930931 Chr.9: 137252919-137252919 on Build GRCh38
rs775092464 Chr.9: 137252924-137252924 on Build GRCh38
rs576486142 Chr.9: 137253039-137253039 on Build GRCh38
rs745460939 Chr.9: 137252916-137252916 on Build GRCh38
rs149372405 Chr.9: 137253035-137253035 on Build GRCh38
rs755157038 Chr.9: 137252897-137252897 on Build GRCh38
rs758473564 Chr.9: 137252983-137252983 on Build GRCh38
rs757711891 Chr.9: 137252999-137252999 on Build GRCh38
rs762529777 Chr.9: 137252925-137252925 on Build GRCh38
rs533959242 Chr.9: 137252933-137252933 on Build GRCh38
rs770332510 Chr.9: 137253012-137253012 on Build GRCh38
rs758518664 Chr.9: 137252911-137252911 on Build GRCh38
rs752473487 Chr.9: 137252881-137252881 on Build GRCh38
rs749005578 Chr.9: 137253054-137253054 on Build GRCh38
rs762815281 Chr.9: 137252896-137252896 on Build GRCh38
rs768222932 Chr.9: 137253061-137253061 on Build GRCh38
rs961821446 Chr.9: 137253007-137253007 on Build GRCh38
rs748246080 Chr.9: 137252909-137252909 on Build GRCh38
rs767274550 Chr.9: 137252943-137252943 on Build GRCh38
rs769364642 Chr.9: 137252922-137252922 on Build GRCh38
rs913923788 Chr.9: 137253024-137253024 on Build GRCh38
rs779822669 Chr.9: 137253052-137253052 on Build GRCh38
rs201688390 Chr.9: 137253001-137253001 on Build GRCh38
rs757070055 Chr.9: 137252893-137252893 on Build GRCh38
rs751587892 Chr.9: 137252892-137252892 on Build GRCh38
rs781009381 Chr.9: 137252940-137252940 on Build GRCh38
rs756140794 Chr.9: 137252924-137252925 on Build GRCh38
rs778225662 Chr.9: 137252912-137252912 on Build GRCh38
rs777891575 Chr.9: 137252990-137252990 on Build GRCh38
rs763763256 Chr.9: 137252932-137252932 on Build GRCh38
rs776876228 Chr.9: 137252915-137252915 on Build GRCh38
rs755176139 Chr.9: 137252963-137252963 on Build GRCh38
rs904222486 Chr.9: 137252908-137252908 on Build GRCh38
rs745612788 Chr.9: 137253016-137253016 on Build GRCh38
rs747539546 Chr.9: 137252913-137252913 on Build GRCh38
rs996242177 Chr.9: 137252910-137252910 on Build GRCh38
rs778920642 Chr.9: 137252901-137252901 on Build GRCh38
rs771317074 Chr.9: 137252914-137252914 on Build GRCh38
rs746241676 Chr.9: 137253005-137253005 on Build GRCh38
rs535888751 Chr.9: 137253062-137253062 on Build GRCh38
rs761668446 Chr.9: 137252942-137252942 on Build GRCh38
rs747495429 Chr.9: 137252995-137252995 on Build GRCh38
rs547276451 Chr.9: 137252944-137252944 on Build GRCh38
rs765419688 Chr.9: 137252968-137252968 on Build GRCh38
rs752952394 Chr.9: 137252980-137252980 on Build GRCh38
rs754032954 Chr.9: 137252894-137252894 on Build GRCh38
rs539927334 Chr.9: 137253021-137253021 on Build GRCh38
rs1043164728 Chr.9: 137252907-137252907 on Build GRCh38
rs769438559 Chr.9: 137252909-137252940 on Build GRCh38

View Full Product Details