Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 142917032-142917295 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP11B2
Entrez Gene ID
1585
Gene Name
cytochrome P450 family 11 subfamily B member 2
Gene Aliases
ALDOS, CPN2, CYP11B, CYP11BL, CYPXIB2, P-450C18, P450C18, P450aldo
UniGene
Hs.632054
Gene Chromosome Location
Chr.8: 142910559-142917843 on Build GRCh38
Transcripts
NM_000498.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3645977 c.333G>A p.M111I Chr.8: 142917121-142917121 on Build GRCh38
5903680 c.245A>G p.N82S Chr.8: 142917209-142917209 on Build GRCh38
3645978 c.291G>A p.V97V Chr.8: 142917163-142917163 on Build GRCh38
6357982 c.344C>T p.P115L Chr.8: 142917110-142917110 on Build GRCh38
4975081 c.365A>T p.H122L Chr.8: 142917089-142917089 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs553894090 Chr.8: 142917245-142917245 on Build GRCh38
rs778434633 Chr.8: 142917250-142917250 on Build GRCh38
rs909005694 Chr.8: 142917131-142917131 on Build GRCh38
rs760842682 Chr.8: 142917137-142917137 on Build GRCh38
rs72554628 Chr.8: 142917208-142917208 on Build GRCh38
rs753318703 Chr.8: 142917184-142917184 on Build GRCh38
rs754516886 Chr.8: 142917249-142917249 on Build GRCh38
rs766593546 Chr.8: 142917230-142917231 on Build GRCh38
rs370785966 Chr.8: 142917173-142917173 on Build GRCh38
rs761120323 Chr.8: 142917226-142917227 on Build GRCh38
rs200283987 Chr.8: 142917198-142917198 on Build GRCh38
rs754372589 Chr.8: 142917232-142917233 on Build GRCh38
rs769098663 Chr.8: 142917121-142917121 on Build GRCh38
rs763745447 Chr.8: 142917081-142917081 on Build GRCh38
rs778618899 Chr.8: 142917194-142917194 on Build GRCh38
rs965343088 Chr.8: 142917065-142917065 on Build GRCh38
rs200788428 Chr.8: 142917055-142917055 on Build GRCh38
rs774948236 Chr.8: 142917058-142917058 on Build GRCh38
rs141789054 Chr.8: 142917172-142917172 on Build GRCh38
rs777017799 Chr.8: 142917149-142917149 on Build GRCh38
rs748879556 Chr.8: 142917119-142917119 on Build GRCh38
rs781165909 Chr.8: 142917086-142917086 on Build GRCh38
rs768833174 Chr.8: 142917053-142917053 on Build GRCh38
rs61757315 Chr.8: 142917261-142917261 on Build GRCh38
rs751360145 Chr.8: 142917166-142917166 on Build GRCh38
rs571318985 Chr.8: 142917230-142917230 on Build GRCh38
rs765466655 Chr.8: 142917163-142917163 on Build GRCh38
rs761513424 Chr.8: 142917238-142917238 on Build GRCh38
rs61757314 Chr.8: 142917224-142917224 on Build GRCh38
rs769635236 Chr.8: 142917223-142917223 on Build GRCh38
rs762727830 Chr.8: 142917150-142917150 on Build GRCh38
rs373048150 Chr.8: 142917195-142917195 on Build GRCh38
rs779683417 Chr.8: 142917112-142917112 on Build GRCh38
rs1040348554 Chr.8: 142917227-142917227 on Build GRCh38
rs768041693 Chr.8: 142917060-142917060 on Build GRCh38
rs963281999 Chr.8: 142917136-142917136 on Build GRCh38
rs538500712 Chr.8: 142917234-142917234 on Build GRCh38
rs778245279 Chr.8: 142917260-142917260 on Build GRCh38
rs752336898 Chr.8: 142917252-142917252 on Build GRCh38
rs751545446 Chr.8: 142917085-142917085 on Build GRCh38
rs771164401 Chr.8: 142917215-142917215 on Build GRCh38
rs754640524 Chr.8: 142917191-142917191 on Build GRCh38
rs372556807 Chr.8: 142917102-142917102 on Build GRCh38
rs781760998 Chr.8: 142917265-142917265 on Build GRCh38
rs769967283 Chr.8: 142917231-142917231 on Build GRCh38
rs761855534 Chr.8: 142917202-142917203 on Build GRCh38
rs146655862 Chr.8: 142917069-142917069 on Build GRCh38
rs758283490 Chr.8: 142917080-142917080 on Build GRCh38
rs202054758 Chr.8: 142917052-142917052 on Build GRCh38
rs768789908 Chr.8: 142917057-142917059 on Build GRCh38
rs776889318 Chr.8: 142917219-142917219 on Build GRCh38
rs767762322 Chr.8: 142917222-142917224 on Build GRCh38
rs199870799 Chr.8: 142917178-142917178 on Build GRCh38
rs771078383 Chr.8: 142917138-142917138 on Build GRCh38
rs527956959 Chr.8: 142917128-142917128 on Build GRCh38
rs773477950 Chr.8: 142917064-142917064 on Build GRCh38
rs774484085 Chr.8: 142917180-142917181 on Build GRCh38
rs779502707 Chr.8: 142917182-142917182 on Build GRCh38
rs567226780 Chr.8: 142917132-142917132 on Build GRCh38
rs769482132 Chr.8: 142917050-142917050 on Build GRCh38
rs755632147 Chr.8: 142917181-142917181 on Build GRCh38
rs147927980 Chr.8: 142917070-142917070 on Build GRCh38
rs549023364 Chr.8: 142917129-142917129 on Build GRCh38
rs745436748 Chr.8: 142917087-142917087 on Build GRCh38
rs754048592 Chr.8: 142917074-142917074 on Build GRCh38
rs753350110 Chr.8: 142917248-142917248 on Build GRCh38
rs13276226 Chr.8: 142917246-142917246 on Build GRCh38
rs758713422 Chr.8: 142917255-142917255 on Build GRCh38

View Full Product Details