Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 68840123-68840396 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EFNB1
Entrez Gene ID
1947
Gene Name
ephrin B1
Gene Aliases
CFND, CFNS, EFB1, EFL3, EPLG2, Elk-L, LERK2
UniGene
Hs.144700
Gene Chromosome Location
Chr.X: 68828997-68842164 on Build GRCh38
Transcripts
NM_004429.4

SNP Detail

SNP ID Chromosome Location
rs1050128606 Chr.X: 68840150-68840150 on Build GRCh38
rs772092082 Chr.X: 68840275-68840275 on Build GRCh38
rs201249059 Chr.X: 68840340-68840340 on Build GRCh38
rs376706341 Chr.X: 68840279-68840279 on Build GRCh38
rs755591908 Chr.X: 68840337-68840337 on Build GRCh38
rs7890214 Chr.X: 68840345-68840345 on Build GRCh38
rs760769932 Chr.X: 68840288-68840288 on Build GRCh38
rs35567709 Chr.X: 68840372-68840372 on Build GRCh38
rs139138728 Chr.X: 68840298-68840298 on Build GRCh38
rs748953203 Chr.X: 68840233-68840233 on Build GRCh38
rs779487954 Chr.X: 68840338-68840338 on Build GRCh38
rs775140374 Chr.X: 68840226-68840226 on Build GRCh38
rs753155032 Chr.X: 68840319-68840319 on Build GRCh38
rs771797163 Chr.X: 68840348-68840348 on Build GRCh38
rs759896368 Chr.X: 68840315-68840315 on Build GRCh38
rs768426785 Chr.X: 68840256-68840256 on Build GRCh38
rs778301964 Chr.X: 68840336-68840336 on Build GRCh38
rs766378296 Chr.X: 68840295-68840295 on Build GRCh38
rs765607277 Chr.X: 68840373-68840373 on Build GRCh38
rs748930955 Chr.X: 68840339-68840339 on Build GRCh38
rs376359844 Chr.X: 68840357-68840357 on Build GRCh38
rs369393911 Chr.X: 68840317-68840317 on Build GRCh38
rs758811344 Chr.X: 68840328-68840328 on Build GRCh38
rs768333182 Chr.X: 68840344-68840344 on Build GRCh38
rs769217553 Chr.X: 68840197-68840197 on Build GRCh38
rs761704495 Chr.X: 68840270-68840270 on Build GRCh38
rs897423032 Chr.X: 68840346-68840346 on Build GRCh38
rs773137160 Chr.X: 68840351-68840351 on Build GRCh38
rs369662868 Chr.X: 68840358-68840358 on Build GRCh38
rs144441935 Chr.X: 68840258-68840258 on Build GRCh38

View Full Product Details