Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 98794605-98794878 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DOCK9
Entrez Gene ID
23348
Gene Name
dedicator of cytokinesis 9
Gene Aliases
ZIZ1, ZIZIMIN1
UniGene
Hs.596105
Gene Chromosome Location
Chr.13: 98793487-99086406 on Build GRCh38
Transcripts
NM_001130048.1 NM_001318849.1 NM_015296.2 XM_005254034.3 XM_005254035.3 XM_006719922.1 XM_006719924.2 XM_006719925.1 XM_006719931.3 XM_006719932.1 XM_006719936.1 XM_006719937.3 XM_017020503.1 XM_017020504.1 XM_017020505.1 XM_017020506.1 XM_017020507.1 XM_017020508.1 XM_017020509.1 XM_017020510.1 XM_017020511.1 XM_017020512.1 XM_017020513.1 XM_017020514.1 XM_017020515.1 XM_017020516.1 XM_017020517.1 XM_017020518.1 XM_017020519.1 XM_017020520.1 XM_017020521.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1952333 c.6267G>A p.S2089S Chr.13: 98794641-98794641 on Build GRCh38
4840513 c.6266C>T p.S2089L Chr.13: 98794642-98794642 on Build GRCh38
5456413 c.6187G>A p.V2063I Chr.13: 98794721-98794721 on Build GRCh38
6648481 c.6203T>G p.L2068R Chr.13: 98794705-98794705 on Build GRCh38
1740032 c.6244C>T p.R2082* Chr.13: 98794714-98794714 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376651254 Chr.13: 98794664-98794664 on Build GRCh38
rs764005812 Chr.13: 98794641-98794641 on Build GRCh38
rs371011966 Chr.13: 98794778-98794778 on Build GRCh38
rs773827419 Chr.13: 98794690-98794690 on Build GRCh38
rs750751006 Chr.13: 98794630-98794630 on Build GRCh38
rs144464149 Chr.13: 98794662-98794662 on Build GRCh38
rs182904172 Chr.13: 98794730-98794730 on Build GRCh38
rs754502168 Chr.13: 98794757-98794757 on Build GRCh38
rs770050776 Chr.13: 98794790-98794790 on Build GRCh38
rs746230011 Chr.13: 98794788-98794788 on Build GRCh38
rs763043197 Chr.13: 98794799-98794799 on Build GRCh38
rs755655293 Chr.13: 98794667-98794667 on Build GRCh38
rs752162002 Chr.13: 98794721-98794721 on Build GRCh38
rs771147223 Chr.13: 98794693-98794693 on Build GRCh38
rs780877117 Chr.13: 98794666-98794666 on Build GRCh38
rs756465455 Chr.13: 98794731-98794733 on Build GRCh38
rs776673614 Chr.13: 98794629-98794629 on Build GRCh38
rs1056572255 Chr.13: 98794796-98794796 on Build GRCh38
rs751404028 Chr.13: 98794655-98794655 on Build GRCh38
rs748796866 Chr.13: 98794685-98794685 on Build GRCh38
rs759679773 Chr.13: 98794631-98794631 on Build GRCh38
rs190124590 Chr.13: 98794726-98794726 on Build GRCh38
rs373217503 Chr.13: 98794711-98794711 on Build GRCh38
rs554564990 Chr.13: 98794647-98794647 on Build GRCh38
rs753442874 Chr.13: 98794752-98794752 on Build GRCh38
rs374223923 Chr.13: 98794743-98794743 on Build GRCh38
rs812808 Chr.13: 98794751-98794751 on Build GRCh38
rs374618395 Chr.13: 98794823-98794823 on Build GRCh38
rs369024723 Chr.13: 98794692-98794692 on Build GRCh38
rs569691232 Chr.13: 98794769-98794769 on Build GRCh38
rs775657930 Chr.13: 98794797-98794797 on Build GRCh38
rs759606820 Chr.13: 98794709-98794709 on Build GRCh38
rs752633742 Chr.13: 98794636-98794636 on Build GRCh38
rs543722179 Chr.13: 98794673-98794673 on Build GRCh38
rs751405068 Chr.13: 98794725-98794725 on Build GRCh38
rs755891748 Chr.13: 98794746-98794746 on Build GRCh38
rs988489161 Chr.13: 98794736-98794736 on Build GRCh38
rs765239941 Chr.13: 98794635-98794635 on Build GRCh38
rs539522315 Chr.13: 98794697-98794697 on Build GRCh38
rs199674506 Chr.13: 98794714-98794714 on Build GRCh38
rs377598866 Chr.13: 98794713-98794713 on Build GRCh38
rs781200040 Chr.13: 98794768-98794768 on Build GRCh38
rs548258424 Chr.13: 98794766-98794766 on Build GRCh38
rs1016953910 Chr.13: 98794640-98794640 on Build GRCh38
rs772759239 Chr.13: 98794689-98794689 on Build GRCh38
rs530847224 Chr.13: 98794844-98794844 on Build GRCh38
rs940183678 Chr.13: 98794810-98794810 on Build GRCh38

View Full Product Details