Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 112785070-112785337 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP11A
Entrez Gene ID
23250
Gene Name
ATPase phospholipid transporting 11A
Gene Aliases
ATPIH, ATPIS
UniGene
Hs.29189
Gene Chromosome Location
Chr.13: 112690034-112887168 on Build GRCh38
Transcripts
NM_015205.2 NM_032189.3 XM_005268299.4 XM_005268300.4 XM_005268301.4 XM_005268303.4 XM_005268305.4 XM_005268306.4 XM_011537480.2 XM_017020490.1 XM_017020491.1 XM_017020492.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
945477 c.115G>A p.A39T Chr.13: 112785210-112785210 on Build GRCh38
6607030 c.63G>A p.V21V Chr.13: 112785158-112785158 on Build GRCh38
6298115 c.162+8G>T p.? Chr.13: 112785265-112785265 on Build GRCh38
6298114 c.162+8G>T p.? Chr.13: 112785265-112785265 on Build GRCh38
4460794 c.117C>T p.A39A Chr.13: 112785212-112785212 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs966293160 Chr.13: 112785313-112785313 on Build GRCh38
rs771483674 Chr.13: 112785185-112785185 on Build GRCh38
rs36001240 Chr.13: 112785111-112785111 on Build GRCh38
rs749804371 Chr.13: 112785184-112785184 on Build GRCh38
rs374875666 Chr.13: 112785177-112785177 on Build GRCh38
rs769716430 Chr.13: 112785204-112785204 on Build GRCh38
rs749139111 Chr.13: 112785300-112785300 on Build GRCh38
rs781312787 Chr.13: 112785159-112785159 on Build GRCh38
rs746539954 Chr.13: 112785199-112785199 on Build GRCh38
rs746334645 Chr.13: 112785256-112785256 on Build GRCh38
rs530661358 Chr.13: 112785241-112785241 on Build GRCh38
rs766294796 Chr.13: 112785213-112785213 on Build GRCh38
rs908132477 Chr.13: 112785232-112785232 on Build GRCh38
rs376312436 Chr.13: 112785233-112785233 on Build GRCh38
rs770116199 Chr.13: 112785091-112785091 on Build GRCh38
rs985922440 Chr.13: 112785113-112785113 on Build GRCh38
rs771571626 Chr.13: 112785102-112785102 on Build GRCh38
rs770740651 Chr.13: 112785301-112785301 on Build GRCh38
rs760216713 Chr.13: 112785110-112785110 on Build GRCh38
rs200309535 Chr.13: 112785175-112785175 on Build GRCh38
rs151175621 Chr.13: 112785287-112785287 on Build GRCh38
rs765075738 Chr.13: 112785122-112785122 on Build GRCh38
rs763013321 Chr.13: 112785308-112785309 on Build GRCh38
rs777387203 Chr.13: 112785139-112785139 on Build GRCh38
rs779467346 Chr.13: 112785188-112785188 on Build GRCh38
rs373090964 Chr.13: 112785132-112785132 on Build GRCh38
rs753156635 Chr.13: 112785203-112785203 on Build GRCh38
rs368654745 Chr.13: 112785119-112785119 on Build GRCh38
rs986016196 Chr.13: 112785307-112785307 on Build GRCh38
rs367618974 Chr.13: 112785265-112785265 on Build GRCh38
rs571264378 Chr.13: 112785314-112785314 on Build GRCh38
rs371369782 Chr.13: 112785270-112785270 on Build GRCh38
rs750416000 Chr.13: 112785129-112785129 on Build GRCh38
rs927464116 Chr.13: 112785242-112785242 on Build GRCh38
rs1041695463 Chr.13: 112785290-112785290 on Build GRCh38
rs755084330 Chr.13: 112785140-112785140 on Build GRCh38
rs772996863 Chr.13: 112785292-112785292 on Build GRCh38
rs751486329 Chr.13: 112785216-112785216 on Build GRCh38
rs570781602 Chr.13: 112785284-112785284 on Build GRCh38
rs774181538 Chr.13: 112785309-112785309 on Build GRCh38
rs202173070 Chr.13: 112785117-112785117 on Build GRCh38
rs11616475 Chr.13: 112785272-112785272 on Build GRCh38
rs769688583 Chr.13: 112785222-112785223 on Build GRCh38
rs778305258 Chr.13: 112785100-112785100 on Build GRCh38
rs546993460 Chr.13: 112785311-112785311 on Build GRCh38
rs776314383 Chr.13: 112785118-112785118 on Build GRCh38
rs368959592 Chr.13: 112785308-112785308 on Build GRCh38
rs148138018 Chr.13: 112785235-112785235 on Build GRCh38
rs935680014 Chr.13: 112785144-112785144 on Build GRCh38
rs1054153652 Chr.13: 112785173-112785173 on Build GRCh38
rs201361256 Chr.13: 112785251-112785251 on Build GRCh38
rs755266989 Chr.13: 112785161-112785161 on Build GRCh38
rs911758226 Chr.13: 112785114-112785114 on Build GRCh38
rs188267977 Chr.13: 112785227-112785227 on Build GRCh38
rs751650645 Chr.13: 112785135-112785135 on Build GRCh38
rs549038564 Chr.13: 112785250-112785250 on Build GRCh38
rs774801520 Chr.13: 112785107-112785107 on Build GRCh38
rs779282744 Chr.13: 112785176-112785176 on Build GRCh38
rs370459187 Chr.13: 112785131-112785131 on Build GRCh38
rs756268096 Chr.13: 112785245-112785245 on Build GRCh38
rs754477254 Chr.13: 112785259-112785259 on Build GRCh38
rs773322179 Chr.13: 112785300-112785300 on Build GRCh38
rs1004276984 Chr.13: 112785214-112785214 on Build GRCh38

View Full Product Details