Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 109488286-109488559 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATXN7L2
Entrez Gene ID
127002
Gene Name
ataxin 7 like 2
Gene Aliases
-
UniGene
Hs.118248
Gene Chromosome Location
Chr.1: 109483904-109492804 on Build GRCh38
Transcripts
NM_153340.4 XM_005270445.3 XM_005270446.2 XM_011540638.2 XM_011540639.2 XM_011540641.2 XM_011540644.2 XM_011540645.1 XM_011540646.2 XM_017000267.1 XM_017000268.1 XM_017000269.1 XM_017000270.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
423402 c.704A>G p.K235R Chr.1: 109488386-109488386 on Build GRCh38
6225612 c.763A>G p.T255A Chr.1: 109488445-109488445 on Build GRCh38
1332454 c.711C>T p.C237C Chr.1: 109488393-109488393 on Build GRCh38
5129127 c.783+5delC p.? Chr.1: 109488470-109488470 on Build GRCh38
893514 c.746A>C p.E249A Chr.1: 109488428-109488428 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs773402570 Chr.1: 109488393-109488393 on Build GRCh38
rs751765463 Chr.1: 109488448-109488448 on Build GRCh38
rs373941223 Chr.1: 109488350-109488350 on Build GRCh38
rs761132662 Chr.1: 109488396-109488396 on Build GRCh38
rs763034206 Chr.1: 109488434-109488434 on Build GRCh38
rs771275433 Chr.1: 109488493-109488493 on Build GRCh38
rs186952013 Chr.1: 109488310-109488310 on Build GRCh38
rs768370571 Chr.1: 109488379-109488379 on Build GRCh38
rs756123694 Chr.1: 109488470-109488470 on Build GRCh38
rs750381199 Chr.1: 109488465-109488465 on Build GRCh38
rs779651345 Chr.1: 109488471-109488471 on Build GRCh38
rs777104777 Chr.1: 109488406-109488406 on Build GRCh38
rs760097795 Chr.1: 109488408-109488408 on Build GRCh38
rs747804817 Chr.1: 109488488-109488488 on Build GRCh38
rs377654845 Chr.1: 109488392-109488392 on Build GRCh38
rs377393957 Chr.1: 109488519-109488519 on Build GRCh38
rs781191079 Chr.1: 109488453-109488453 on Build GRCh38
rs940411123 Chr.1: 109488445-109488445 on Build GRCh38
rs369423678 Chr.1: 109488450-109488450 on Build GRCh38
rs765299259 Chr.1: 109488415-109488415 on Build GRCh38
rs1027167067 Chr.1: 109488449-109488449 on Build GRCh38
rs752829351 Chr.1: 109488417-109488417 on Build GRCh38
rs769414525 Chr.1: 109488363-109488363 on Build GRCh38
rs1144593 Chr.1: 109488323-109488323 on Build GRCh38
rs191730623 Chr.1: 109488534-109488534 on Build GRCh38
rs745848656 Chr.1: 109488356-109488356 on Build GRCh38
rs374140650 Chr.1: 109488486-109488486 on Build GRCh38
rs766770957 Chr.1: 109488405-109488405 on Build GRCh38
rs1008470736 Chr.1: 109488317-109488317 on Build GRCh38
rs778596661 Chr.1: 109488487-109488487 on Build GRCh38
rs758226004 Chr.1: 109488470-109488470 on Build GRCh38
rs1006834272 Chr.1: 109488504-109488504 on Build GRCh38
rs745638279 Chr.1: 109488337-109488339 on Build GRCh38
rs899978443 Chr.1: 109488340-109488340 on Build GRCh38
rs907628779 Chr.1: 109488432-109488432 on Build GRCh38
rs552714197 Chr.1: 109488318-109488318 on Build GRCh38
rs764405873 Chr.1: 109488447-109488447 on Build GRCh38
rs1014565045 Chr.1: 109488526-109488526 on Build GRCh38
rs200333729 Chr.1: 109488460-109488460 on Build GRCh38
rs748854536 Chr.1: 109488472-109488472 on Build GRCh38
rs748824855 Chr.1: 109488376-109488376 on Build GRCh38
rs996106359 Chr.1: 109488354-109488354 on Build GRCh38

View Full Product Details