Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 64804790-64805049 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MEN1
Entrez Gene ID
4221
Gene Name
menin 1
Gene Aliases
MEAI, SCG2
UniGene
Hs.423348
Gene Chromosome Location
Chr.11: 64803514-64811294 on Build GRCh38
Transcripts
NM_000244.3 NM_130799.2 NM_130800.2 NM_130801.2 NM_130802.2 NM_130803.2 NM_130804.2 XM_005274001.4 XM_011545040.1 XM_011545041.2 XM_011545042.2 XM_017017765.1 XM_017017766.1 XM_017017767.1 XM_017017768.1 XM_017017769.1 XM_017017770.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
23011 c.1361_1362delAG p.K454fs*76 Chr.11: 64804805-64804806 on Build GRCh38
23026 c.1351-4C>T p.? Chr.11: 64804820-64804820 on Build GRCh38
2164473 c.1367G>A p.R456H Chr.11: 64804800-64804800 on Build GRCh38
4607519 c.1342G>T p.E448* Chr.11: 64805042-64805042 on Build GRCh38
1237074 c.1351-1_1357delGGTGCGGC p.? Chr.11: 64804810-64804817 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753219137 Chr.11: 64804829-64804830 on Build GRCh38
rs751932519 Chr.11: 64804824-64804824 on Build GRCh38
rs758957568 Chr.11: 64804856-64804862 on Build GRCh38
rs761444487 Chr.11: 64804944-64804944 on Build GRCh38
rs977303482 Chr.11: 64804839-64804839 on Build GRCh38
rs757569943 Chr.11: 64804826-64804826 on Build GRCh38
rs753431102 Chr.11: 64804837-64804837 on Build GRCh38
rs377038984 Chr.11: 64804858-64804858 on Build GRCh38
rs991260111 Chr.11: 64804965-64804965 on Build GRCh38
rs654440 Chr.11: 64804931-64804931 on Build GRCh38
rs863224810 Chr.11: 64804813-64804813 on Build GRCh38
rs764570645 Chr.11: 64805022-64805023 on Build GRCh38
rs746828409 Chr.11: 64804985-64804985 on Build GRCh38
rs746077651 Chr.11: 64805028-64805028 on Build GRCh38
rs539720166 Chr.11: 64804994-64804994 on Build GRCh38
rs754539993 Chr.11: 64804843-64804843 on Build GRCh38
rs777011577 Chr.11: 64805008-64805008 on Build GRCh38
rs775922507 Chr.11: 64804812-64804812 on Build GRCh38
rs777519719 Chr.11: 64804984-64804984 on Build GRCh38
rs770778266 Chr.11: 64804999-64804999 on Build GRCh38
rs758331652 Chr.11: 64804866-64804866 on Build GRCh38
rs778479960 Chr.11: 64804844-64804844 on Build GRCh38
rs555673667 Chr.11: 64804981-64804981 on Build GRCh38
rs763514855 Chr.11: 64804815-64804815 on Build GRCh38
rs938500157 Chr.11: 64804945-64804945 on Build GRCh38
rs794728629 Chr.11: 64804817-64804817 on Build GRCh38
rs954426009 Chr.11: 64804903-64804903 on Build GRCh38
rs794728644 Chr.11: 64805023-64805033 on Build GRCh38
rs915659869 Chr.11: 64804978-64804978 on Build GRCh38
rs766052368 Chr.11: 64804832-64804832 on Build GRCh38
rs765943168 Chr.11: 64804826-64804826 on Build GRCh38
rs764408631 Chr.11: 64804820-64804820 on Build GRCh38

View Full Product Details