Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 46218196-46218449 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPARA
Entrez Gene ID
5465
Gene Name
peroxisome proliferator activated receptor alpha
Gene Aliases
NR1C1, PPAR, PPARalpha, hPPAR
UniGene
Hs.103110
Gene Chromosome Location
Chr.22: 46150534-46243756 on Build GRCh38
Transcripts
NM_001001928.2 NM_005036.4 XM_005261655.3 XM_005261656.3 XM_006724269.3 XM_006724270.3 XM_011530239.2 XM_011530240.2 XM_011530241.2 XM_011530242.2 XM_011530243.2 XM_011530244.2 XM_011530245.2 XM_017028839.1 XM_017028840.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4432096 c.443A>G p.K148R Chr.22: 46218336-46218336 on Build GRCh38
3713713 c.444G>C p.K148N Chr.22: 46218337-46218337 on Build GRCh38
4878551 c.493G>A p.G165R Chr.22: 46218386-46218386 on Build GRCh38
6038554 c.508G>A p.A170T Chr.22: 46218401-46218401 on Build GRCh38
5018709 c.373T>A p.F125I Chr.22: 46218266-46218266 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs549840611 Chr.22: 46218245-46218245 on Build GRCh38
rs1800204 Chr.22: 46218273-46218273 on Build GRCh38
rs755862696 Chr.22: 46218236-46218236 on Build GRCh38
rs939270657 Chr.22: 46218401-46218401 on Build GRCh38
rs766370613 Chr.22: 46218314-46218314 on Build GRCh38
rs768647025 Chr.22: 46218361-46218361 on Build GRCh38
rs1022063762 Chr.22: 46218357-46218357 on Build GRCh38
rs565902106 Chr.22: 46218290-46218290 on Build GRCh38
rs780342579 Chr.22: 46218275-46218275 on Build GRCh38
rs147652884 Chr.22: 46218310-46218310 on Build GRCh38
rs1800206 Chr.22: 46218377-46218377 on Build GRCh38
rs113379388 Chr.22: 46218311-46218311 on Build GRCh38
rs200450634 Chr.22: 46218316-46218316 on Build GRCh38
rs781405965 Chr.22: 46218363-46218363 on Build GRCh38
rs779436126 Chr.22: 46218332-46218332 on Build GRCh38
rs752332877 Chr.22: 46218235-46218235 on Build GRCh38
rs779053935 Chr.22: 46218385-46218385 on Build GRCh38
rs201467784 Chr.22: 46218246-46218246 on Build GRCh38
rs889086763 Chr.22: 46218276-46218276 on Build GRCh38
rs758977679 Chr.22: 46218223-46218223 on Build GRCh38
rs146427416 Chr.22: 46218400-46218400 on Build GRCh38
rs4253752 Chr.22: 46218241-46218242 on Build GRCh38
rs1036892981 Chr.22: 46218255-46218255 on Build GRCh38
rs6413511 Chr.22: 46218250-46218250 on Build GRCh38
rs17847852 Chr.22: 46218218-46218218 on Build GRCh38
rs759527309 Chr.22: 46218296-46218296 on Build GRCh38
rs767028190 Chr.22: 46218231-46218231 on Build GRCh38
rs747200358 Chr.22: 46218279-46218279 on Build GRCh38
rs1031173136 Chr.22: 46218419-46218419 on Build GRCh38
rs748330466 Chr.22: 46218370-46218370 on Build GRCh38
rs557774897 Chr.22: 46218315-46218315 on Build GRCh38
rs549236046 Chr.22: 46218285-46218285 on Build GRCh38
rs759044698 Chr.22: 46218336-46218336 on Build GRCh38
rs777968163 Chr.22: 46218243-46218243 on Build GRCh38
rs577158678 Chr.22: 46218328-46218328 on Build GRCh38
rs750956786 Chr.22: 46218333-46218333 on Build GRCh38
rs780254708 Chr.22: 46218340-46218340 on Build GRCh38
rs747058496 Chr.22: 46218349-46218349 on Build GRCh38
rs768967658 Chr.22: 46218280-46218280 on Build GRCh38
rs772047921 Chr.22: 46218426-46218426 on Build GRCh38
rs201964347 Chr.22: 46218292-46218292 on Build GRCh38

View Full Product Details