Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 47789416-47789643 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HDAC7
Entrez Gene ID
51564
Gene Name
histone deacetylase 7
Gene Aliases
HD7, HD7A, HDAC7A
UniGene
Hs.200063
Gene Chromosome Location
Chr.12: 47782710-47820612 on Build GRCh38
Transcripts
NM_001098416.3 NM_001308090.1 NM_015401.4 XM_011538478.1 XM_011538479.1 XM_011538480.1 XM_011538481.1 XM_011538482.1 XM_017019455.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6531506 c.2126A>G p.H709R Chr.12: 47789544-47789544 on Build GRCh38
4042123 c.1993C>T p.R665W Chr.12: 47789560-47789560 on Build GRCh38
5913388 c.2116C>T p.P706S Chr.12: 47789554-47789554 on Build GRCh38
5913389 c.1999C>T p.P667S Chr.12: 47789554-47789554 on Build GRCh38
6531507 c.2009A>G p.H670R Chr.12: 47789544-47789544 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747050885 Chr.12: 47789613-47789613 on Build GRCh38
rs770010093 Chr.12: 47789509-47789510 on Build GRCh38
rs775653638 Chr.12: 47789460-47789460 on Build GRCh38
rs780416721 Chr.12: 47789520-47789520 on Build GRCh38
rs766771434 Chr.12: 47789439-47789439 on Build GRCh38
rs200966208 Chr.12: 47789528-47789528 on Build GRCh38
rs538262078 Chr.12: 47789451-47789451 on Build GRCh38
rs369660030 Chr.12: 47789570-47789570 on Build GRCh38
rs765030024 Chr.12: 47789479-47789479 on Build GRCh38
rs751193453 Chr.12: 47789494-47789494 on Build GRCh38
rs748689356 Chr.12: 47789490-47789490 on Build GRCh38
rs772356335 Chr.12: 47789585-47789585 on Build GRCh38
rs568551956 Chr.12: 47789483-47789483 on Build GRCh38
rs941590756 Chr.12: 47789488-47789488 on Build GRCh38
rs755369860 Chr.12: 47789529-47789529 on Build GRCh38
rs1034118749 Chr.12: 47789441-47789441 on Build GRCh38
rs756702943 Chr.12: 47789516-47789516 on Build GRCh38
rs749844923 Chr.12: 47789502-47789502 on Build GRCh38
rs959500048 Chr.12: 47789458-47789458 on Build GRCh38
rs554732825 Chr.12: 47789471-47789471 on Build GRCh38
rs761413521 Chr.12: 47789497-47789497 on Build GRCh38
rs146834304 Chr.12: 47789549-47789549 on Build GRCh38
rs375303475 Chr.12: 47789482-47789482 on Build GRCh38
rs759524611 Chr.12: 47789478-47789478 on Build GRCh38
rs778016514 Chr.12: 47789598-47789598 on Build GRCh38
rs200014024 Chr.12: 47789546-47789546 on Build GRCh38
rs763778723 Chr.12: 47789487-47789487 on Build GRCh38
rs371466541 Chr.12: 47789543-47789543 on Build GRCh38
rs767018105 Chr.12: 47789498-47789498 on Build GRCh38
rs770117832 Chr.12: 47789474-47789474 on Build GRCh38
rs200040296 Chr.12: 47789490-47789490 on Build GRCh38
rs368845512 Chr.12: 47789489-47789489 on Build GRCh38

View Full Product Details