Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 73009722-73009965 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
P4HA1
Entrez Gene ID
5033
Gene Name
prolyl 4-hydroxylase subunit alpha 1
Gene Aliases
P4HA
UniGene
Hs.500047
Gene Chromosome Location
Chr.10: 73007217-73096974 on Build GRCh38
Transcripts
NM_000917.3 NM_001017962.2 NM_001142595.1 NM_001142596.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6905100 c.1528A>T p.K510* Chr.10: 73009813-73009813 on Build GRCh38
3440363 c.1453T>C p.W485R Chr.10: 73009888-73009888 on Build GRCh38
7341543 c.1534+1G>A p.? Chr.10: 73009806-73009806 on Build GRCh38
1168703 c.1495C>T p.R499W Chr.10: 73009846-73009846 on Build GRCh38
6905099 c.1528A>T p.K510* Chr.10: 73009813-73009813 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1013272414 Chr.10: 73009863-73009863 on Build GRCh38
rs762704174 Chr.10: 73009907-73009907 on Build GRCh38
rs751480625 Chr.10: 73009910-73009910 on Build GRCh38
rs201618967 Chr.10: 73009857-73009857 on Build GRCh38
rs200529312 Chr.10: 73009846-73009846 on Build GRCh38
rs199578638 Chr.10: 73009785-73009785 on Build GRCh38
rs759476786 Chr.10: 73009914-73009914 on Build GRCh38
rs779277588 Chr.10: 73009829-73009829 on Build GRCh38
rs748796556 Chr.10: 73009844-73009844 on Build GRCh38
rs777956348 Chr.10: 73009790-73009790 on Build GRCh38
rs182054109 Chr.10: 73009899-73009899 on Build GRCh38
rs757385706 Chr.10: 73009942-73009942 on Build GRCh38
rs779056009 Chr.10: 73009943-73009943 on Build GRCh38
rs78135024 Chr.10: 73009906-73009906 on Build GRCh38
rs563063202 Chr.10: 73009935-73009935 on Build GRCh38
rs761299532 Chr.10: 73009860-73009860 on Build GRCh38
rs776062341 Chr.10: 73009855-73009855 on Build GRCh38
rs760138660 Chr.10: 73009923-73009924 on Build GRCh38
rs766146091 Chr.10: 73009908-73009908 on Build GRCh38
rs377262769 Chr.10: 73009811-73009811 on Build GRCh38
rs749521770 Chr.10: 73009817-73009817 on Build GRCh38
rs746261950 Chr.10: 73009833-73009833 on Build GRCh38
rs201334110 Chr.10: 73009784-73009784 on Build GRCh38
rs200568046 Chr.10: 73009758-73009758 on Build GRCh38
rs200745198 Chr.10: 73009821-73009821 on Build GRCh38
rs34845894 Chr.10: 73009826-73009826 on Build GRCh38
rs767138765 Chr.10: 73009760-73009769 on Build GRCh38
rs764936176 Chr.10: 73009870-73009870 on Build GRCh38
rs775810604 Chr.10: 73009927-73009927 on Build GRCh38
rs750242998 Chr.10: 73009766-73009766 on Build GRCh38
rs950428468 Chr.10: 73009917-73009917 on Build GRCh38
rs765029598 Chr.10: 73009762-73009762 on Build GRCh38
rs199836353 Chr.10: 73009845-73009845 on Build GRCh38
rs200640345 Chr.10: 73009933-73009933 on Build GRCh38

View Full Product Details