Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 125885122-125885395 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NCOA7
Entrez Gene ID
135112
Gene Name
nuclear receptor coactivator 7
Gene Aliases
ERAP140, ESNA1, NCOA7-AS, Nbla00052, Nbla10993, TLDC4, dJ187J11.3
UniGene
Hs.171426
Gene Chromosome Location
Chr.6: 125780948-125932030 on Build GRCh38
Transcripts
NM_001122842.2 NM_001199619.1 NM_001199620.1 NM_001199621.1 NM_181782.4 XM_005266822.4 XM_006715340.3 XM_011535455.2 XM_017010269.1 XM_017010270.1 XM_017010271.1 XM_017010272.1 XM_017010273.1 XM_017010274.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4950400 c.712G>A p.G238S Chr.6: 125885171-125885171 on Build GRCh38
739331 c.785G>T p.G262V Chr.6: 125885244-125885244 on Build GRCh38
3828971 c.810C>T p.C270C Chr.6: 125885269-125885269 on Build GRCh38
739332 c.700G>T p.G234C Chr.6: 125885159-125885159 on Build GRCh38
1072918 c.758A>C p.K253T Chr.6: 125885217-125885217 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770343876 Chr.6: 125885290-125885290 on Build GRCh38
rs773880953 Chr.6: 125885153-125885153 on Build GRCh38
rs773584572 Chr.6: 125885334-125885334 on Build GRCh38
rs564180167 Chr.6: 125885304-125885304 on Build GRCh38
rs759713494 Chr.6: 125885362-125885362 on Build GRCh38
rs762943973 Chr.6: 125885198-125885198 on Build GRCh38
rs960137943 Chr.6: 125885367-125885367 on Build GRCh38
rs764540367 Chr.6: 125885170-125885170 on Build GRCh38
rs144802599 Chr.6: 125885332-125885332 on Build GRCh38
rs1008752090 Chr.6: 125885164-125885164 on Build GRCh38
rs765879974 Chr.6: 125885182-125885182 on Build GRCh38
rs779607503 Chr.6: 125885351-125885354 on Build GRCh38
rs572278982 Chr.6: 125885295-125885295 on Build GRCh38
rs149399962 Chr.6: 125885296-125885296 on Build GRCh38
rs748766933 Chr.6: 125885275-125885275 on Build GRCh38
rs767747792 Chr.6: 125885369-125885369 on Build GRCh38
rs750470281 Chr.6: 125885196-125885196 on Build GRCh38
rs149187270 Chr.6: 125885272-125885272 on Build GRCh38
rs762382381 Chr.6: 125885178-125885178 on Build GRCh38
rs574558612 Chr.6: 125885251-125885251 on Build GRCh38
rs762294306 Chr.6: 125885318-125885318 on Build GRCh38
rs777189437 Chr.6: 125885172-125885172 on Build GRCh38
rs754141471 Chr.6: 125885346-125885346 on Build GRCh38
rs761853674 Chr.6: 125885370-125885370 on Build GRCh38
rs931099663 Chr.6: 125885309-125885309 on Build GRCh38
rs200191767 Chr.6: 125885155-125885155 on Build GRCh38
rs543364532 Chr.6: 125885340-125885340 on Build GRCh38
rs763569841 Chr.6: 125885335-125885335 on Build GRCh38
rs777587206 Chr.6: 125885273-125885273 on Build GRCh38
rs755242256 Chr.6: 125885237-125885237 on Build GRCh38
rs535307523 Chr.6: 125885270-125885270 on Build GRCh38
rs201538729 Chr.6: 125885228-125885228 on Build GRCh38

View Full Product Details