Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 35424630-35424879 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPARD
Entrez Gene ID
5467
Gene Name
peroxisome proliferator activated receptor delta
Gene Aliases
FAAR, NR1C2, NUC1, NUCI, NUCII, PPARB
UniGene
Hs.696032
Gene Chromosome Location
Chr.6: 35342558-35428191 on Build GRCh38
Transcripts
NM_001171818.1 NM_001171819.1 NM_001171820.1 NM_006238.4 NM_177435.2 XM_005249193.1 XM_006715120.1 XM_006715123.1 XM_011514707.1 XM_011514709.1 XM_011514710.1 XM_017010971.1 XM_017010972.1 XM_017010973.1 XM_017010974.1 XM_017010975.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3626001 c.1069C>T p.L357L Chr.6: 35424770-35424770 on Build GRCh38
396199 c.961C>T p.R321C Chr.6: 35424662-35424662 on Build GRCh38
236205 c.1045G>A p.A349T Chr.6: 35424746-35424746 on Build GRCh38
3626000 c.1053C>T p.F351F Chr.6: 35424754-35424754 on Build GRCh38
217380 c.1030G>C p.D344H Chr.6: 35424731-35424731 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199532192 Chr.6: 35424823-35424823 on Build GRCh38
rs772540082 Chr.6: 35424717-35424717 on Build GRCh38
rs974238749 Chr.6: 35424821-35424821 on Build GRCh38
rs760315803 Chr.6: 35424661-35424661 on Build GRCh38
rs200533476 Chr.6: 35424668-35424668 on Build GRCh38
rs747826964 Chr.6: 35424712-35424712 on Build GRCh38
rs756560599 Chr.6: 35424654-35424654 on Build GRCh38
rs189344916 Chr.6: 35424699-35424699 on Build GRCh38
rs201991879 Chr.6: 35424842-35424842 on Build GRCh38
rs775359832 Chr.6: 35424755-35424755 on Build GRCh38
rs755334161 Chr.6: 35424832-35424832 on Build GRCh38
rs139511972 Chr.6: 35424760-35424760 on Build GRCh38
rs796552370 Chr.6: 35424843-35424843 on Build GRCh38
rs771764757 Chr.6: 35424715-35424715 on Build GRCh38
rs868445344 Chr.6: 35424666-35424666 on Build GRCh38
rs766967189 Chr.6: 35424746-35424747 on Build GRCh38
rs767799861 Chr.6: 35424799-35424799 on Build GRCh38
rs201614080 Chr.6: 35424691-35424691 on Build GRCh38
rs776751609 Chr.6: 35424737-35424737 on Build GRCh38
rs755381306 Chr.6: 35424706-35424706 on Build GRCh38
rs1029262828 Chr.6: 35424751-35424751 on Build GRCh38
rs180784946 Chr.6: 35424801-35424801 on Build GRCh38
rs200961108 Chr.6: 35424718-35424718 on Build GRCh38
rs200910320 Chr.6: 35424791-35424791 on Build GRCh38
rs765007268 Chr.6: 35424754-35424754 on Build GRCh38
rs199957473 Chr.6: 35424844-35424844 on Build GRCh38
rs537830530 Chr.6: 35424838-35424838 on Build GRCh38
rs144386007 Chr.6: 35424824-35424824 on Build GRCh38
rs889344882 Chr.6: 35424759-35424759 on Build GRCh38
rs1010114172 Chr.6: 35424716-35424716 on Build GRCh38
rs750529579 Chr.6: 35424653-35424653 on Build GRCh38
rs763362626 Chr.6: 35424812-35424812 on Build GRCh38
rs756223563 Chr.6: 35424820-35424820 on Build GRCh38
rs199828289 Chr.6: 35424761-35424761 on Build GRCh38
rs770953928 Chr.6: 35424719-35424719 on Build GRCh38
rs200150550 Chr.6: 35424727-35424727 on Build GRCh38
rs202146453 Chr.6: 35424656-35424656 on Build GRCh38
rs759390931 Chr.6: 35424746-35424746 on Build GRCh38

View Full Product Details