Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 76403545-76403806 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IQGAP2
Entrez Gene ID
10788
Gene Name
IQ motif containing GTPase activating protein 2
Gene Aliases
-
UniGene
Hs.291030
Gene Chromosome Location
Chr.5: 76403255-76708132 on Build GRCh38
Transcripts
NM_006633.3 XM_017008960.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5420745 c.46+7G>T p.? Chr.5: 76403598-76403598 on Build GRCh38
6361726 c.26T>A p.L9Q Chr.5: 76403571-76403571 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs567282358 Chr.5: 76403653-76403653 on Build GRCh38
rs923598704 Chr.5: 76403702-76403702 on Build GRCh38
rs960851121 Chr.5: 76403766-76403774 on Build GRCh38
rs764579261 Chr.5: 76403632-76403632 on Build GRCh38
rs936335868 Chr.5: 76403704-76403704 on Build GRCh38
rs538497714 Chr.5: 76403761-76403761 on Build GRCh38
rs953883811 Chr.5: 76403634-76403634 on Build GRCh38
rs1022141109 Chr.5: 76403678-76403678 on Build GRCh38
rs759149825 Chr.5: 76403611-76403611 on Build GRCh38
rs892114659 Chr.5: 76403667-76403671 on Build GRCh38
rs368715165 Chr.5: 76403622-76403622 on Build GRCh38
rs569312379 Chr.5: 76403579-76403579 on Build GRCh38
rs754088569 Chr.5: 76403583-76403583 on Build GRCh38
rs187353476 Chr.5: 76403705-76403705 on Build GRCh38
rs912483550 Chr.5: 76403669-76403669 on Build GRCh38
rs919359237 Chr.5: 76403775-76403775 on Build GRCh38
rs374375138 Chr.5: 76403717-76403717 on Build GRCh38
rs747881615 Chr.5: 76403565-76403565 on Build GRCh38
rs930601944 Chr.5: 76403783-76403783 on Build GRCh38
rs916315176 Chr.5: 76403715-76403715 on Build GRCh38
rs62363210 Chr.5: 76403774-76403774 on Build GRCh38
rs906779820 Chr.5: 76403748-76403748 on Build GRCh38
rs140739362 Chr.5: 76403606-76403606 on Build GRCh38
rs940919181 Chr.5: 76403768-76403768 on Build GRCh38
rs572795945 Chr.5: 76403746-76403746 on Build GRCh38
rs906755313 Chr.5: 76403580-76403580 on Build GRCh38
rs763338122 Chr.5: 76403630-76403630 on Build GRCh38
rs771859775 Chr.5: 76403572-76403572 on Build GRCh38
rs770664994 Chr.5: 76403584-76403584 on Build GRCh38
rs535828837 Chr.5: 76403674-76403674 on Build GRCh38
rs1046534031 Chr.5: 76403741-76403741 on Build GRCh38
rs985279089 Chr.5: 76403658-76403658 on Build GRCh38
rs978165625 Chr.5: 76403697-76403697 on Build GRCh38
rs765646425 Chr.5: 76403613-76403613 on Build GRCh38
rs866148017 Chr.5: 76403582-76403582 on Build GRCh38
rs902050569 Chr.5: 76403608-76403608 on Build GRCh38
rs1034849968 Chr.5: 76403716-76403716 on Build GRCh38

View Full Product Details