Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 156557256-156557509 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPD2
Entrez Gene ID
2820
Gene Name
glycerol-3-phosphate dehydrogenase 2
Gene Aliases
GDH2, GPDM, mGPDH
UniGene
Hs.512382
Gene Chromosome Location
Chr.2: 156435290-156586403 on Build GRCh38
Transcripts
NM_000408.4 NM_001083112.2 XM_005246469.2 XM_011510977.2 XM_011510978.2 XM_011510980.2 XM_017003830.1 XM_017003831.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1007988 c.1086A>G p.T362T Chr.2: 156557503-156557503 on Build GRCh38
5697525 c.1029C>T p.F343F Chr.2: 156557446-156557446 on Build GRCh38
3299648 c.1056T>C p.A352A Chr.2: 156557473-156557473 on Build GRCh38
6848709 c.1016G>A p.R339Q Chr.2: 156557433-156557433 on Build GRCh38
375699 c.1058G>T p.G353V Chr.2: 156557475-156557475 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756980310 Chr.2: 156557419-156557419 on Build GRCh38
rs745525242 Chr.2: 156557440-156557440 on Build GRCh38
rs773373104 Chr.2: 156557380-156557380 on Build GRCh38
rs890736993 Chr.2: 156557396-156557396 on Build GRCh38
rs766451062 Chr.2: 156557350-156557350 on Build GRCh38
rs747504715 Chr.2: 156557470-156557470 on Build GRCh38
rs908811772 Chr.2: 156557395-156557395 on Build GRCh38
rs766373400 Chr.2: 156557327-156557327 on Build GRCh38
rs371381240 Chr.2: 156557467-156557467 on Build GRCh38
rs771535457 Chr.2: 156557466-156557466 on Build GRCh38
rs759376048 Chr.2: 156557433-156557433 on Build GRCh38
rs374133957 Chr.2: 156557483-156557483 on Build GRCh38
rs775190532 Chr.2: 156557471-156557471 on Build GRCh38
rs983085135 Chr.2: 156557325-156557325 on Build GRCh38
rs377620746 Chr.2: 156557418-156557418 on Build GRCh38
rs1017782580 Chr.2: 156557421-156557421 on Build GRCh38
rs1010988379 Chr.2: 156557432-156557432 on Build GRCh38
rs374100828 Chr.2: 156557379-156557379 on Build GRCh38
rs764790881 Chr.2: 156557296-156557296 on Build GRCh38
rs777739698 Chr.2: 156557404-156557404 on Build GRCh38
rs369671755 Chr.2: 156557349-156557349 on Build GRCh38
rs928944281 Chr.2: 156557324-156557324 on Build GRCh38
rs146086957 Chr.2: 156557429-156557429 on Build GRCh38

View Full Product Details