Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 65190629-65190902 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AK4
Entrez Gene ID
205
Gene Name
adenylate kinase 4
Gene Aliases
AK 4, AK3, AK3L1, AK3L2
UniGene
Hs.10862
Gene Chromosome Location
Chr.1: 65147342-65232145 on Build GRCh38
Transcripts
NM_001005353.2 NM_013410.3 NM_203464.2 XM_017000612.1 XM_017000613.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7111805 c.175G>C p.E59Q Chr.1: 65190739-65190739 on Build GRCh38
4476794 c.209C>G p.T70R Chr.1: 65190773-65190773 on Build GRCh38
6597345 c.147A>T p.E49D Chr.1: 65190711-65190711 on Build GRCh38
426591 c.262G>A p.D88N Chr.1: 65190826-65190826 on Build GRCh38
83910 c.226G>A p.E76K Chr.1: 65190790-65190790 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761550887 Chr.1: 65190802-65190802 on Build GRCh38
rs758802413 Chr.1: 65190856-65190856 on Build GRCh38
rs771129711 Chr.1: 65190781-65190781 on Build GRCh38
rs201095991 Chr.1: 65190745-65190745 on Build GRCh38
rs372965063 Chr.1: 65190853-65190853 on Build GRCh38
rs753526076 Chr.1: 65190694-65190694 on Build GRCh38
rs769838095 Chr.1: 65190790-65190790 on Build GRCh38
rs925939551 Chr.1: 65190687-65190687 on Build GRCh38
rs149952312 Chr.1: 65190751-65190751 on Build GRCh38
rs199575222 Chr.1: 65190793-65190793 on Build GRCh38
rs778643434 Chr.1: 65190732-65190732 on Build GRCh38
rs765447936 Chr.1: 65190840-65190841 on Build GRCh38
rs765899833 Chr.1: 65190844-65190844 on Build GRCh38
rs781183664 Chr.1: 65190784-65190784 on Build GRCh38
rs778347805 Chr.1: 65190865-65190865 on Build GRCh38
rs760676184 Chr.1: 65190830-65190830 on Build GRCh38
rs771906783 Chr.1: 65190805-65190805 on Build GRCh38
rs760158861 Chr.1: 65190682-65190682 on Build GRCh38
rs370616612 Chr.1: 65190729-65190729 on Build GRCh38
rs367641986 Chr.1: 65190851-65190851 on Build GRCh38
rs765917651 Chr.1: 65190693-65190693 on Build GRCh38
rs750384864 Chr.1: 65190670-65190670 on Build GRCh38
rs750597215 Chr.1: 65190842-65190843 on Build GRCh38
rs929965584 Chr.1: 65190775-65190775 on Build GRCh38
rs374922038 Chr.1: 65190806-65190806 on Build GRCh38
rs745970024 Chr.1: 65190789-65190789 on Build GRCh38
rs143331044 Chr.1: 65190776-65190776 on Build GRCh38
rs752939193 Chr.1: 65190734-65190734 on Build GRCh38
rs753229457 Chr.1: 65190846-65190846 on Build GRCh38
rs1005262135 Chr.1: 65190848-65190848 on Build GRCh38
rs752441127 Chr.1: 65190854-65190854 on Build GRCh38

View Full Product Details