Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 89001363-89001636 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
S1PR3
Entrez Gene ID
1903
Gene Name
sphingosine-1-phosphate receptor 3
Gene Aliases
EDG-3, EDG3, LPB3, S1P3
UniGene
Hs.585118
Gene Chromosome Location
Chr.9: 88991409-89005155 on Build GRCh38
Transcripts
NM_005226.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3908734 c.371C>T p.A124V Chr.9: 89001571-89001571 on Build GRCh38
487649 c.428T>C p.M143T Chr.9: 89001628-89001628 on Build GRCh38
6116677 c.250T>C p.C84R Chr.9: 89001450-89001450 on Build GRCh38
3908735 c.397G>A p.A133T Chr.9: 89001597-89001597 on Build GRCh38
3849045 c.328G>A p.V110I Chr.9: 89001528-89001528 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs546398741 Chr.9: 89001418-89001418 on Build GRCh38
rs765103971 Chr.9: 89001456-89001456 on Build GRCh38
rs774529851 Chr.9: 89001540-89001540 on Build GRCh38
rs771513659 Chr.9: 89001538-89001538 on Build GRCh38
rs146344787 Chr.9: 89001569-89001569 on Build GRCh38
rs138828961 Chr.9: 89001455-89001455 on Build GRCh38
rs144659554 Chr.9: 89001436-89001436 on Build GRCh38
rs1001633903 Chr.9: 89001477-89001477 on Build GRCh38
rs754000616 Chr.9: 89001598-89001598 on Build GRCh38
rs533101623 Chr.9: 89001592-89001592 on Build GRCh38
rs966639811 Chr.9: 89001533-89001533 on Build GRCh38
rs749723383 Chr.9: 89001537-89001537 on Build GRCh38
rs999390790 Chr.9: 89001559-89001559 on Build GRCh38
rs867697242 Chr.9: 89001466-89001466 on Build GRCh38
rs868073895 Chr.9: 89001484-89001484 on Build GRCh38
rs1056242367 Chr.9: 89001479-89001479 on Build GRCh38
rs746847329 Chr.9: 89001503-89001503 on Build GRCh38
rs944178717 Chr.9: 89001416-89001416 on Build GRCh38
rs138547551 Chr.9: 89001440-89001440 on Build GRCh38
rs201536511 Chr.9: 89001526-89001526 on Build GRCh38
rs541053919 Chr.9: 89001587-89001587 on Build GRCh38
rs758401590 Chr.9: 89001497-89001497 on Build GRCh38
rs752549324 Chr.9: 89001464-89001464 on Build GRCh38
rs758220931 Chr.9: 89001602-89001602 on Build GRCh38
rs766646598 Chr.9: 89001452-89001452 on Build GRCh38
rs778851016 Chr.9: 89001404-89001404 on Build GRCh38
rs199731931 Chr.9: 89001471-89001471 on Build GRCh38
rs759653844 Chr.9: 89001463-89001463 on Build GRCh38
rs781097409 Chr.9: 89001389-89001389 on Build GRCh38
rs371960454 Chr.9: 89001578-89001578 on Build GRCh38
rs780998699 Chr.9: 89001518-89001518 on Build GRCh38
rs777888427 Chr.9: 89001490-89001490 on Build GRCh38
rs763570069 Chr.9: 89001451-89001451 on Build GRCh38
rs751247905 Chr.9: 89001607-89001607 on Build GRCh38
rs768405364 Chr.9: 89001432-89001432 on Build GRCh38
rs757136729 Chr.9: 89001403-89001403 on Build GRCh38
rs768158498 Chr.9: 89001509-89001509 on Build GRCh38
rs750358039 Chr.9: 89001496-89001496 on Build GRCh38
rs564201526 Chr.9: 89001591-89001591 on Build GRCh38
rs766373530 Chr.9: 89001606-89001606 on Build GRCh38
rs779968450 Chr.9: 89001419-89001419 on Build GRCh38
rs199610835 Chr.9: 89001465-89001465 on Build GRCh38
rs752823423 Chr.9: 89001392-89001392 on Build GRCh38
rs772921383 Chr.9: 89001527-89001527 on Build GRCh38
rs776236082 Chr.9: 89001435-89001435 on Build GRCh38
rs756188776 Chr.9: 89001476-89001476 on Build GRCh38
rs773872325 Chr.9: 89001441-89001441 on Build GRCh38
rs759975385 Chr.9: 89001552-89001552 on Build GRCh38
rs1053495537 Chr.9: 89001445-89001445 on Build GRCh38
rs767748061 Chr.9: 89001467-89001467 on Build GRCh38
rs114214031 Chr.9: 89001500-89001500 on Build GRCh38
rs746611494 Chr.9: 89001425-89001425 on Build GRCh38
rs776379928 Chr.9: 89001415-89001415 on Build GRCh38
rs373882187 Chr.9: 89001513-89001513 on Build GRCh38
rs747607283 Chr.9: 89001522-89001522 on Build GRCh38

View Full Product Details