Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 4938518-4938765 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC25A11
Entrez Gene ID
8402
Gene Name
solute carrier family 25 member 11
Gene Aliases
OGC, SLC20A4
UniGene
Hs.184877
Gene Chromosome Location
Chr.17: 4937130-4940167 on Build GRCh38
Transcripts
NM_001165417.1 NM_001165418.1 NM_003562.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
164372 c.520G>C p.E174Q Chr.17: 4938538-4938538 on Build GRCh38
6780493 c.517C>T p.R173W Chr.17: 4938541-4938541 on Build GRCh38
4067586 c.508C>T p.R170* Chr.17: 4938550-4938550 on Build GRCh38
4726781 c.498C>T p.N166N Chr.17: 4938560-4938560 on Build GRCh38
6219344 c.456-2delA p.? Chr.17: 4938604-4938604 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770093144 Chr.17: 4938560-4938560 on Build GRCh38
rs531311106 Chr.17: 4938659-4938659 on Build GRCh38
rs181186410 Chr.17: 4938711-4938711 on Build GRCh38
rs771053325 Chr.17: 4938595-4938595 on Build GRCh38
rs1052534231 Chr.17: 4938720-4938720 on Build GRCh38
rs969879111 Chr.17: 4938690-4938690 on Build GRCh38
rs1027843953 Chr.17: 4938582-4938582 on Build GRCh38
rs749256407 Chr.17: 4938585-4938585 on Build GRCh38
rs371448058 Chr.17: 4938586-4938586 on Build GRCh38
rs1005165997 Chr.17: 4938556-4938556 on Build GRCh38
rs762312430 Chr.17: 4938636-4938636 on Build GRCh38
rs935477602 Chr.17: 4938703-4938703 on Build GRCh38
rs568132585 Chr.17: 4938727-4938727 on Build GRCh38
rs771056182 Chr.17: 4938709-4938709 on Build GRCh38
rs139352273 Chr.17: 4938597-4938597 on Build GRCh38
rs752127372 Chr.17: 4938541-4938541 on Build GRCh38
rs748570174 Chr.17: 4938554-4938554 on Build GRCh38
rs922784900 Chr.17: 4938699-4938700 on Build GRCh38
rs764759812 Chr.17: 4938624-4938624 on Build GRCh38
rs777795729 Chr.17: 4938737-4938737 on Build GRCh38
rs776390927 Chr.17: 4938610-4938610 on Build GRCh38
rs1045475762 Chr.17: 4938734-4938734 on Build GRCh38
rs755602409 Chr.17: 4938542-4938542 on Build GRCh38
rs992177525 Chr.17: 4938658-4938658 on Build GRCh38
rs769850413 Chr.17: 4938672-4938672 on Build GRCh38
rs570161931 Chr.17: 4938694-4938694 on Build GRCh38
rs552414858 Chr.17: 4938613-4938613 on Build GRCh38
rs1023684994 Chr.17: 4938663-4938663 on Build GRCh38
rs760598504 Chr.17: 4938605-4938605 on Build GRCh38
rs756502252 Chr.17: 4938735-4938735 on Build GRCh38
rs768623748 Chr.17: 4938608-4938608 on Build GRCh38
rs372521210 Chr.17: 4938540-4938540 on Build GRCh38
rs749510276 Chr.17: 4938738-4938738 on Build GRCh38
rs781661516 Chr.17: 4938543-4938543 on Build GRCh38
rs189215876 Chr.17: 4938644-4938644 on Build GRCh38
rs773346094 Chr.17: 4938695-4938695 on Build GRCh38
rs753069610 Chr.17: 4938728-4938728 on Build GRCh38
rs775110172 Chr.17: 4938559-4938559 on Build GRCh38
rs375769507 Chr.17: 4938566-4938566 on Build GRCh38

View Full Product Details