Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 49110683-49110944 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
USP19
Entrez Gene ID
10869
Gene Name
ubiquitin specific peptidase 19
Gene Aliases
ZMYND9
UniGene
Hs.255596
Gene Chromosome Location
Chr.3: 49108046-49120938 on Build GRCh38
Transcripts
NM_001199160.1 NM_001199161.1 NM_001199162.1 NM_006677.2 XM_005264823.2 XM_005264824.2 XM_005264825.2 XM_005264826.2 XM_005264827.2 XM_005264829.2 XM_005264830.2 XM_005264831.2 XM_006712946.2 XM_006712947.2 XM_006712948.2 XM_006712949.2 XM_006712950.2 XM_006712951.2 XM_006712952.2 XM_006712953.1 XM_017005615.1 XM_017005616.1 XM_017005617.1 XM_017005618.1 XM_017005619.1 XM_017005620.1 XM_017005621.1 XM_017005622.1 XM_017005623.1 XM_017005624.1 XM_017005625.1 XM_017005626.1 XM_017005627.1 XM_017005628.1 XM_017005629.1 XM_017005630.1 XM_017005631.1 XM_017005632.1 XM_017005633.1 XM_017005634.1 XM_017005635.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5509928 c.3656G>A p.R1219H Chr.3: 49110747-49110747 on Build GRCh38
6201505 c.3578A>T p.K1193M Chr.3: 49110825-49110825 on Build GRCh38
6480450 c.3565C>T p.R1189C Chr.3: 49110838-49110838 on Build GRCh38
1485310 c.3615C>A p.I1205I Chr.3: 49110788-49110788 on Build GRCh38
2849420 c.3601C>T p.P1201S Chr.3: 49110802-49110802 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773907250 Chr.3: 49110851-49110851 on Build GRCh38
rs762371255 Chr.3: 49110890-49110890 on Build GRCh38
rs759118522 Chr.3: 49110875-49110875 on Build GRCh38
rs748032466 Chr.3: 49110712-49110712 on Build GRCh38
rs758698553 Chr.3: 49110797-49110797 on Build GRCh38
rs371746052 Chr.3: 49110884-49110884 on Build GRCh38
rs748724669 Chr.3: 49110838-49110838 on Build GRCh38
rs761567911 Chr.3: 49110883-49110883 on Build GRCh38
rs1050596905 Chr.3: 49110735-49110735 on Build GRCh38
rs761049999 Chr.3: 49110762-49110762 on Build GRCh38
rs35826923 Chr.3: 49110716-49110717 on Build GRCh38
rs146952729 Chr.3: 49110791-49110791 on Build GRCh38
rs777384080 Chr.3: 49110818-49110818 on Build GRCh38
rs772585093 Chr.3: 49110748-49110748 on Build GRCh38
rs746760816 Chr.3: 49110704-49110704 on Build GRCh38
rs201609350 Chr.3: 49110781-49110781 on Build GRCh38
rs563967011 Chr.3: 49110787-49110787 on Build GRCh38
rs753768562 Chr.3: 49110875-49110876 on Build GRCh38
rs751768909 Chr.3: 49110802-49110802 on Build GRCh38
rs377178527 Chr.3: 49110921-49110921 on Build GRCh38
rs768525390 Chr.3: 49110706-49110706 on Build GRCh38
rs770456455 Chr.3: 49110757-49110757 on Build GRCh38
rs781313579 Chr.3: 49110707-49110707 on Build GRCh38
rs765356241 Chr.3: 49110788-49110788 on Build GRCh38
rs769018302 Chr.3: 49110879-49110879 on Build GRCh38
rs775970938 Chr.3: 49110759-49110759 on Build GRCh38
rs762246359 Chr.3: 49110785-49110785 on Build GRCh38
rs769443042 Chr.3: 49110722-49110722 on Build GRCh38
rs748155148 Chr.3: 49110812-49110812 on Build GRCh38
rs552145464 Chr.3: 49110893-49110893 on Build GRCh38
rs781076406 Chr.3: 49110808-49110808 on Build GRCh38
rs780164404 Chr.3: 49110799-49110799 on Build GRCh38
rs770425871 Chr.3: 49110846-49110846 on Build GRCh38
rs769164646 Chr.3: 49110906-49110906 on Build GRCh38
rs897766997 Chr.3: 49110823-49110823 on Build GRCh38
rs764401082 Chr.3: 49110770-49110770 on Build GRCh38
rs1013418977 Chr.3: 49110880-49110880 on Build GRCh38
rs762977359 Chr.3: 49110920-49110920 on Build GRCh38
rs754829256 Chr.3: 49110805-49110805 on Build GRCh38
rs368798901 Chr.3: 49110703-49110703 on Build GRCh38

View Full Product Details