Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 83385195-83385468 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SEMA3E
Entrez Gene ID
9723
Gene Name
semaphorin 3E
Gene Aliases
M-SEMAH, M-SemaK, SEMAH, coll-5
UniGene
Hs.528721
Gene Chromosome Location
Chr.7: 83363906-83649163 on Build GRCh38
Transcripts
NM_001178129.1 NM_012431.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7187095 c.1740T>A p.D580E Chr.7: 83385429-83385429 on Build GRCh38
1643386 c.1856G>A p.R619H Chr.7: 83385313-83385313 on Build GRCh38
6012760 c.1736-10C>T p.? Chr.7: 83385443-83385443 on Build GRCh38
3663364 c.1790G>A p.S597N Chr.7: 83385379-83385379 on Build GRCh38
346005 c.1736-2A>G p.? Chr.7: 83385435-83385435 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756256536 Chr.7: 83385255-83385255 on Build GRCh38
rs781452966 Chr.7: 83385249-83385249 on Build GRCh38
rs753040834 Chr.7: 83385409-83385409 on Build GRCh38
rs774915686 Chr.7: 83385397-83385397 on Build GRCh38
rs746987765 Chr.7: 83385360-83385360 on Build GRCh38
rs60017278 Chr.7: 83385234-83385234 on Build GRCh38
rs140160399 Chr.7: 83385313-83385313 on Build GRCh38
rs371092782 Chr.7: 83385363-83385363 on Build GRCh38
rs560983031 Chr.7: 83385335-83385335 on Build GRCh38
rs576576885 Chr.7: 83385419-83385419 on Build GRCh38
rs756346342 Chr.7: 83385342-83385342 on Build GRCh38
rs749388864 Chr.7: 83385260-83385260 on Build GRCh38
rs761147897 Chr.7: 83385415-83385415 on Build GRCh38
rs143701381 Chr.7: 83385428-83385428 on Build GRCh38
rs772875017 Chr.7: 83385384-83385384 on Build GRCh38
rs143631464 Chr.7: 83385314-83385314 on Build GRCh38
rs762503685 Chr.7: 83385386-83385386 on Build GRCh38
rs201699077 Chr.7: 83385431-83385431 on Build GRCh38
rs1038193638 Chr.7: 83385299-83385299 on Build GRCh38
rs778055435 Chr.7: 83385258-83385258 on Build GRCh38
rs148962198 Chr.7: 83385366-83385366 on Build GRCh38
rs759912253 Chr.7: 83385320-83385320 on Build GRCh38
rs754158945 Chr.7: 83385430-83385430 on Build GRCh38
rs771975608 Chr.7: 83385263-83385263 on Build GRCh38
rs776265763 Chr.7: 83385280-83385280 on Build GRCh38
rs971383920 Chr.7: 83385376-83385376 on Build GRCh38
rs377608971 Chr.7: 83385433-83385433 on Build GRCh38
rs950606105 Chr.7: 83385235-83385235 on Build GRCh38
rs778141353 Chr.7: 83385343-83385343 on Build GRCh38
rs750533810 Chr.7: 83385439-83385439 on Build GRCh38
rs529914797 Chr.7: 83385341-83385341 on Build GRCh38
rs768082404 Chr.7: 83385408-83385408 on Build GRCh38
rs927545176 Chr.7: 83385315-83385315 on Build GRCh38
rs762483567 Chr.7: 83385312-83385312 on Build GRCh38
rs375687801 Chr.7: 83385424-83385424 on Build GRCh38
rs145707650 Chr.7: 83385355-83385355 on Build GRCh38
rs373351427 Chr.7: 83385275-83385275 on Build GRCh38
rs202040246 Chr.7: 83385278-83385278 on Build GRCh38
rs772643898 Chr.7: 83385303-83385303 on Build GRCh38
rs868825060 Chr.7: 83385383-83385383 on Build GRCh38
rs200744403 Chr.7: 83385250-83385250 on Build GRCh38
rs769453523 Chr.7: 83385297-83385297 on Build GRCh38
rs755322226 Chr.7: 83385244-83385244 on Build GRCh38
rs377026121 Chr.7: 83385347-83385347 on Build GRCh38
rs769396568 Chr.7: 83385370-83385370 on Build GRCh38
rs761514237 Chr.7: 83385285-83385285 on Build GRCh38
rs760019484 Chr.7: 83385400-83385400 on Build GRCh38

View Full Product Details