Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 1488313-1488584 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PTX4
Entrez Gene ID
390667
Gene Name
pentraxin 4
Gene Aliases
C16orf38
UniGene
Hs.553770
Gene Chromosome Location
Chr.16: 1485939-1488467 on Build GRCh38
Transcripts
NM_001013658.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6760306 c.76G>A p.G26R Chr.16: 1488392-1488392 on Build GRCh38
4748521 c.56G>A p.R19Q Chr.16: 1488412-1488412 on Build GRCh38
3957152 c.111G>A p.L37L Chr.16: 1488357-1488357 on Build GRCh38
3728977 c.126+3G>A p.? Chr.16: 1488339-1488339 on Build GRCh38
5452061 c.126+1G>A p.? Chr.16: 1488341-1488341 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs61743346 Chr.16: 1488343-1488343 on Build GRCh38
rs751163469 Chr.16: 1488365-1488365 on Build GRCh38
rs766152448 Chr.16: 1488346-1488346 on Build GRCh38
rs199725289 Chr.16: 1488355-1488355 on Build GRCh38
rs577050680 Chr.16: 1488375-1488375 on Build GRCh38
rs747252825 Chr.16: 1488391-1488391 on Build GRCh38
rs368853793 Chr.16: 1488498-1488498 on Build GRCh38
rs753236158 Chr.16: 1488428-1488428 on Build GRCh38
rs766127812 Chr.16: 1488478-1488486 on Build GRCh38
rs2745103 Chr.16: 1488463-1488463 on Build GRCh38
rs760037419 Chr.16: 1488467-1488467 on Build GRCh38
rs779522056 Chr.16: 1488383-1488383 on Build GRCh38
rs773889436 Chr.16: 1488461-1488461 on Build GRCh38
rs372706858 Chr.16: 1488452-1488452 on Build GRCh38
rs753186680 Chr.16: 1488478-1488478 on Build GRCh38
rs762768499 Chr.16: 1488398-1488421 on Build GRCh38
rs765186585 Chr.16: 1488395-1488395 on Build GRCh38
rs575639679 Chr.16: 1488392-1488392 on Build GRCh38
rs34993742 Chr.16: 1488559-1488559 on Build GRCh38
rs968278638 Chr.16: 1488446-1488446 on Build GRCh38
rs2745102 Chr.16: 1488432-1488432 on Build GRCh38
rs141533929 Chr.16: 1488367-1488367 on Build GRCh38
rs767282592 Chr.16: 1488465-1488465 on Build GRCh38
rs781347378 Chr.16: 1488447-1488447 on Build GRCh38
rs763643714 Chr.16: 1488404-1488404 on Build GRCh38
rs200690297 Chr.16: 1488405-1488405 on Build GRCh38
rs149153276 Chr.16: 1488424-1488424 on Build GRCh38
rs745663189 Chr.16: 1488449-1488449 on Build GRCh38
rs778097156 Chr.16: 1488485-1488485 on Build GRCh38
rs373294212 Chr.16: 1488466-1488466 on Build GRCh38
rs766899912 Chr.16: 1488413-1488413 on Build GRCh38
rs143253362 Chr.16: 1488517-1488517 on Build GRCh38
rs754285813 Chr.16: 1488418-1488418 on Build GRCh38
rs756492378 Chr.16: 1488373-1488373 on Build GRCh38
rs772134706 Chr.16: 1488387-1488387 on Build GRCh38
rs542290749 Chr.16: 1488488-1488488 on Build GRCh38
rs759441725 Chr.16: 1488349-1488349 on Build GRCh38
rs756939030 Chr.16: 1488412-1488412 on Build GRCh38
rs542121480 Chr.16: 1488421-1488421 on Build GRCh38
rs145194704 Chr.16: 1488393-1488393 on Build GRCh38
rs769640852 Chr.16: 1488502-1488502 on Build GRCh38
rs773419314 Chr.16: 1488341-1488341 on Build GRCh38
rs976766943 Chr.16: 1488552-1488553 on Build GRCh38
rs776619689 Chr.16: 1488347-1488347 on Build GRCh38
rs747248914 Chr.16: 1488436-1488436 on Build GRCh38
rs757882025 Chr.16: 1488358-1488358 on Build GRCh38
rs751543228 Chr.16: 1488501-1488502 on Build GRCh38
rs748181421 Chr.16: 1488386-1488386 on Build GRCh38
rs773366012 Chr.16: 1488390-1488390 on Build GRCh38
rs780559425 Chr.16: 1488374-1488374 on Build GRCh38
rs771169048 Chr.16: 1488444-1488444 on Build GRCh38
rs1022821291 Chr.16: 1488445-1488445 on Build GRCh38
rs758476855 Chr.16: 1488431-1488431 on Build GRCh38
rs772214290 Chr.16: 1488339-1488339 on Build GRCh38
rs150414885 Chr.16: 1488378-1488378 on Build GRCh38
rs751674308 Chr.16: 1488486-1488486 on Build GRCh38
rs374769106 Chr.16: 1488477-1488477 on Build GRCh38
rs762379909 Chr.16: 1488398-1488398 on Build GRCh38
rs895378001 Chr.16: 1488371-1488371 on Build GRCh38
rs946788252 Chr.16: 1488342-1488342 on Build GRCh38
rs111506404 Chr.16: 1488459-1488459 on Build GRCh38
rs1040499 Chr.16: 1488362-1488362 on Build GRCh38
rs137991965 Chr.16: 1488456-1488456 on Build GRCh38
rs762941522 Chr.16: 1488458-1488458 on Build GRCh38
rs559417572 Chr.16: 1488394-1488394 on Build GRCh38
rs376991299 Chr.16: 1488384-1488384 on Build GRCh38
rs552443178 Chr.16: 1488354-1488354 on Build GRCh38
rs373471949 Chr.16: 1488484-1488484 on Build GRCh38
rs765631851 Chr.16: 1488472-1488472 on Build GRCh38
rs916843966 Chr.16: 1488473-1488473 on Build GRCh38
rs758919885 Chr.16: 1488481-1488481 on Build GRCh38
rs746074625 Chr.16: 1488501-1488501 on Build GRCh38

View Full Product Details