Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 125895388-125895631 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HYLS1
Entrez Gene ID
219844
Gene Name
HYLS1, centriolar and ciliogenesis associated
Gene Aliases
HLS
UniGene
Hs.98133
Gene Chromosome Location
Chr.11: 125883614-125900646 on Build GRCh38
Transcripts
NM_001134793.1 NM_145014.2 XM_005271430.2 XM_006718777.3 XM_011542659.2 XM_017017320.1 XM_017017321.1 NM_001271985.1 NM_031307.3 XM_005271687.3 XM_005271688.3

Gene Symbol
PUS3
Entrez Gene ID
83480
Gene Name
pseudouridylate synthase 3
Gene Aliases
2610020J05Rik, FKSG32
UniGene
Hs.660922
Gene Chromosome Location
Chr.11: 125893485-125903774 on Build GRCh38
Transcripts
NM_001134793.1 NM_145014.2 XM_005271430.2 XM_006718777.3 XM_011542659.2 XM_017017320.1 XM_017017321.1 NM_001271985.1 NM_031307.3 XM_005271687.3 XM_005271688.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4165691 c.760G>T p.G254C Chr.11: 125895408-125895408 on Build GRCh38
3383298 c.712C>G p.Q238E Chr.11: 125895456-125895456 on Build GRCh38
6848250 c.588delC p.Y197fs*8 Chr.11: 125895580-125895580 on Build GRCh38
686490 c.601C>T p.R201C Chr.11: 125895567-125895567 on Build GRCh38
401298 c.748G>A p.G250S Chr.11: 125895420-125895420 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs533170206 Chr.11: 125895580-125895580 on Build GRCh38
rs780046582 Chr.11: 125895599-125895599 on Build GRCh38
rs763863295 Chr.11: 125895423-125895423 on Build GRCh38
rs781140167 Chr.11: 125895451-125895451 on Build GRCh38
rs564112431 Chr.11: 125895566-125895566 on Build GRCh38
rs140780584 Chr.11: 125895472-125895472 on Build GRCh38
rs746287849 Chr.11: 125895577-125895577 on Build GRCh38
rs372488492 Chr.11: 125895567-125895567 on Build GRCh38
rs148017767 Chr.11: 125895468-125895468 on Build GRCh38
rs775846693 Chr.11: 125895489-125895489 on Build GRCh38
rs757155405 Chr.11: 125895435-125895435 on Build GRCh38
rs750180726 Chr.11: 125895517-125895517 on Build GRCh38
rs761503592 Chr.11: 125895513-125895513 on Build GRCh38
rs150075002 Chr.11: 125895484-125895484 on Build GRCh38
rs1048501650 Chr.11: 125895455-125895455 on Build GRCh38
rs549981261 Chr.11: 125895581-125895581 on Build GRCh38
rs189235230 Chr.11: 125895563-125895563 on Build GRCh38
rs367946467 Chr.11: 125895508-125895508 on Build GRCh38
rs576405108 Chr.11: 125895590-125895590 on Build GRCh38
rs930064187 Chr.11: 125895452-125895452 on Build GRCh38
rs779262413 Chr.11: 125895570-125895570 on Build GRCh38
rs751448230 Chr.11: 125895557-125895557 on Build GRCh38
rs755733425 Chr.11: 125895412-125895412 on Build GRCh38
rs763478603 Chr.11: 125895509-125895509 on Build GRCh38
rs377076175 Chr.11: 125895591-125895591 on Build GRCh38
rs752183515 Chr.11: 125895407-125895407 on Build GRCh38
rs750088950 Chr.11: 125895598-125895598 on Build GRCh38
rs774649580 Chr.11: 125895473-125895473 on Build GRCh38
rs1007332055 Chr.11: 125895485-125895485 on Build GRCh38
rs370110541 Chr.11: 125895582-125895582 on Build GRCh38
rs75995371 Chr.11: 125895487-125895487 on Build GRCh38
rs780100387 Chr.11: 125895550-125895550 on Build GRCh38
rs758427069 Chr.11: 125895535-125895535 on Build GRCh38
rs778825745 Chr.11: 125895439-125895439 on Build GRCh38
rs745818046 Chr.11: 125895442-125895442 on Build GRCh38
rs775961025 Chr.11: 125895585-125895585 on Build GRCh38
rs753425638 Chr.11: 125895434-125895434 on Build GRCh38
rs773552329 Chr.11: 125895471-125895471 on Build GRCh38
rs765159522 Chr.11: 125895514-125895514 on Build GRCh38
rs376187060 Chr.11: 125895464-125895464 on Build GRCh38
rs866552678 Chr.11: 125895419-125895419 on Build GRCh38
rs758375656 Chr.11: 125895447-125895447 on Build GRCh38

View Full Product Details