Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 19878430-19878703 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF253
Entrez Gene ID
56242
Gene Name
zinc finger protein 253
Gene Aliases
BMZF-1, BMZF1, ZNF411
UniGene
Hs.301059
Gene Chromosome Location
Chr.19: 19865905-19893484 on Build GRCh38
Transcripts
NM_021047.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5855171 c.121G>A p.V41I Chr.19: 19878598-19878598 on Build GRCh38
5855172 c.121G>A p.V41I Chr.19: 19878598-19878598 on Build GRCh38
286772 c.34G>T p.E12* Chr.19: 19878511-19878511 on Build GRCh38
6264369 c.130+8C>T p.? Chr.19: 19878615-19878615 on Build GRCh38
6291524 c.118T>C p.L40L Chr.19: 19878595-19878595 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755484158 Chr.19: 19878554-19878554 on Build GRCh38
rs886160364 Chr.19: 19878485-19878485 on Build GRCh38
rs1047217882 Chr.19: 19878650-19878650 on Build GRCh38
rs766171578 Chr.19: 19878544-19878544 on Build GRCh38
rs190030835 Chr.19: 19878487-19878487 on Build GRCh38
rs767406118 Chr.19: 19878535-19878535 on Build GRCh38
rs753613453 Chr.19: 19878553-19878553 on Build GRCh38
rs763061966 Chr.19: 19878505-19878505 on Build GRCh38
rs898583222 Chr.19: 19878517-19878517 on Build GRCh38
rs201876433 Chr.19: 19878633-19878633 on Build GRCh38
rs775503342 Chr.19: 19878501-19878501 on Build GRCh38
rs771509649 Chr.19: 19878590-19878590 on Build GRCh38
rs747660045 Chr.19: 19878572-19878572 on Build GRCh38
rs778596289 Chr.19: 19878567-19878567 on Build GRCh38
rs746231637 Chr.19: 19878615-19878615 on Build GRCh38
rs10406883 Chr.19: 19878486-19878486 on Build GRCh38
rs574877794 Chr.19: 19878463-19878463 on Build GRCh38
rs750399985 Chr.19: 19878536-19878536 on Build GRCh38
rs775764232 Chr.19: 19878627-19878627 on Build GRCh38
rs777193750 Chr.19: 19878599-19878599 on Build GRCh38
rs770185224 Chr.19: 19878616-19878616 on Build GRCh38
rs778036763 Chr.19: 19878576-19878577 on Build GRCh38
rs747556846 Chr.19: 19878576-19878576 on Build GRCh38
rs564010866 Chr.19: 19878587-19878587 on Build GRCh38
rs757945607 Chr.19: 19878575-19878575 on Build GRCh38
rs771452536 Chr.19: 19878470-19878470 on Build GRCh38
rs546133308 Chr.19: 19878644-19878644 on Build GRCh38
rs765542664 Chr.19: 19878488-19878488 on Build GRCh38
rs372446403 Chr.19: 19878559-19878559 on Build GRCh38
rs900434925 Chr.19: 19878491-19878491 on Build GRCh38
rs60758402 Chr.19: 19878473-19878474 on Build GRCh38
rs754689149 Chr.19: 19878561-19878561 on Build GRCh38
rs755975940 Chr.19: 19878538-19878538 on Build GRCh38
rs532139029 Chr.19: 19878654-19878654 on Build GRCh38
rs916563956 Chr.19: 19878473-19878473 on Build GRCh38

View Full Product Details